ContactUsLinkComponent
CPC® Miniature Quick-Disconnect Fittings, Panel-Mount Hose Barb Bodies
CPC® Miniature Quick-Disconnect Fittings, Panel-Mount Hose Barb Bodies
Certificates
undefined
CPC® Miniature Quick-Disconnect Fittings, Panel-Mount Hose Barb Bodies
Specifications
  • Max. temperature:
  • 82 °C (180 °F)
  • 82 °C (180 °F)
  • 82 °C (180 °F)
  • 82 °C (180 °F)
  • Min. temperature:
  • 0 °C (32 °F)
  • 0 °C (32 °F)
  • –40 °C (–40 °F)
  • –40 °C (–40 °F)
  • O-Ring material:
  • EPDM
  • EPDM
  • Buna N
  • Buna N
  • Sterilisation:
  • Ethylene oxide (ETO) or gamma
  • Ethylene oxide (ETO) or gamma
  • Autoclave or ethylene oxide (ETO)
  • Autoclave or ethylene oxide (ETO)
  • Valve material:
  • None
  • None
  • None
  • None
  • Warranty:
  • 3 Months
  • 1 year
  • 3 Months
  • 3 Months