ContactUsLinkComponent
Parker Hannifin Compression Fitting Inserts for Soft Tubing
Parker Hannifin P-P-6 Insert for Soft Tubing, White PP, 3/8" OD x 1/4" ID; 10/Pk
Catalog # MFLX06391-30
undefined
Parker Hannifin P-P-6 Insert for Soft Tubing, White PP, 3/8" OD x 1/4" ID; 10/Pk
Catalog # MFLX06391-30
Specifications
  • Barb size/tubing int.Ø (port 2):
    ¼"
  • Connection Type:
    Compression Insert
  • Flow pattern:
    Straight
  • Description:
    P-P-6 Insert for Soft Tubing, White PP, ⅜" OD×¼" ID; 10/Pk
  • Sterilisation:
    Ethylene oxide (ETO) only
  • Pk:
    10
  • Fitting material:
    Polypropylene
Specifications
About this item
Allows compression fittings to be used with soft tubing.
  • Secure your flexible, soft tubing
Compression fittings are an excellent choice to connect rigid tubing systems and withstand higher pressures than barbed fittings. Assemble the fitting to the tubing by sliding the tubing through a sealing ferrule and using the nut to compress down to form a secure seal.
Inserts fit in the end of the soft tubing to provide the rigidity required for adequate compression and a proper seal.
  • Barb size/tubing int.Ø (port 2):
    ¼"
  • Connection Type:
    Compression Insert
  • Flow pattern:
    Straight
  • Description:
    P-P-6 Insert for Soft Tubing, White PP, ⅜" OD×¼" ID; 10/Pk
  • Environmentally Preferable:
  • Sterilisation:
    Ethylene oxide (ETO) only
  • Pk:
    10
  • Fitting material:
    Polypropylene