Masterflex® Plastic Assorted Fitting Kits, Avantor®
About this item
Get organized today!
- Ideal for general-purpose and single-use applications, to connect tubing with rest of system
- Determine which connector type is needed for your application by contacting free technical application assistance*
- Global market leader in range of materials for most extensive chemical compatibility
- Single-use custom configuration solutions to minimise downtime
- Regulatory approvals to match your unique application
- Assorted fittings come in an impact-resistant plastic box with secure snap-shut latches to prevent spillage
- Labeled compartments let you easily locate the fittings you want
- Order storage racks separately
Organize your workstation with these convenient fitting kits. Assorted fittings come in impact resistant plastic box with secure latch to keep fittings organized and in place. Label compartments make fitting location quick and easy. Choose from four materials: high-density polyethylene (HDPE), black nylon, natural polypropylene, and natural PVDF.
26 Options Available Below
Product Details & Documents
Get organized today!
- Ideal for general-purpose and single-use applications, to connect tubing with rest of system
- Determine which connector type is needed for your application by contacting free technical application assistance*
- Global market leader in range of materials for most extensive chemical compatibility
- Single-use custom configuration solutions to minimise downtime
- Regulatory approvals to match your unique application
- Assorted fittings come in an impact-resistant plastic box with secure snap-shut latches to prevent spillage
- Labeled compartments let you easily locate the fittings you want
- Order storage racks separately
Organize your workstation with these convenient fitting kits. Assorted fittings come in impact resistant plastic box with secure latch to keep fittings organized and in place. Label compartments make fitting location quick and easy. Choose from four materials: high-density polyethylene (HDPE), black nylon, natural polypropylene, and natural PVDF.
Certificates
Search by Lot NumberSpecifications for Product Family
Specifications
- Catalog No:
- MFLX31514-01
- MFLX31514-02
- MFLX31514-05
- MFLX31514-06
- MFLX31514-09
- MFLX31514-10
- MFLX31514-13
- MFLX31514-14
- MFLX31514-17
- MFLX31514-18
- MFLX31514-22
- MFLX31514-25
- MFLX31514-26
- MFLX31514-29
- MFLX31514-32
- MFLX31514-33
- MFLX31514-34
- MFLX41514-03
- MFLX41514-07
- MFLX41514-11
- MFLX41514-15
- MFLX41514-19
- MFLX41514-23
- MFLX41514-27
- MFLX41514-30
- MFLX41514-35
- Model:
- 31514-01
- 31514-02
- 31514-05
- 31514-06
- 31514-09
- 31514-10
- 31514-13
- 31514-14
- 31514-17
- 31514-18
- 31514-22
- 31514-25
- 31514-26
- 31514-29
- 31514-32
- 31514-33
- 31514-34
- 41514-03
- 41514-07
- 41514-11
- 41514-15
- 41514-19
- 41514-23
- 41514-27
- 41514-30
- 41514-35
- Max. temperature:
- 87 °C (190 °F)
- 93 °C (200 °F)
- 87 °C (190 °F)
- 93 °C (200 °F)
- 87 °C (190 °F)
- 93 °C (200 °F)
- 87 °C (190 °F)
- 93 °C (200 °F)
- 87 °C (190 °F)
- 93 °C (200 °F)
- 93 °C (200 °F)
- 87 °C (190 °F)
- 93 °C (200 °F)
- 93 °C (200 °F)
- 87 °C (190 °F)
- 87 °C (190 °F)
- 93 °C (200 °F)
- 135 °C (275 °F)
- 135 °C (275 °F)
- 135 °C (275 °F)
- 135 °C (275 °F)
- 135 °C (275 °F)
- 135 °C (275 °F)
- 135 °C (275 °F)
- 135 °C (275 °F)
- 135 °C (275 °F)
- Min. temperature:
- –53 °C (–65 °F)
- –40 °C (–40 °F)
- –53 °C (–65 °F)
- –40 °C (–40 °F)
- –53 °C (–65 °F)
- –40 °C (–40 °F)
- –53 °C (–65 °F)
- –40 °C (–40 °F)
- –53 °C (–65 °F)
- –40 °C (–40 °F)
- –40 °C (–40 °F)
- –53 °C (–65 °F)
- –40 °C (–40 °F)
- –40 °C (–40 °F)
- –53 °C (–65 °F)
- –53 °C (–65 °F)
- –40 °C (–40 °F)
- –17 °C (0 °F)
- –17 °C (0 °F)
- –17 °C (0 °F)
- –17 °C (0 °F)
- –17 °C (0 °F)
- –17 °C (0 °F)
- –17 °C (0 °F)
- –17 °C (0 °F)
- –17 °C (0 °F)
- Warranty:
- 1 Month
- 1 Month
- 1 Month
- 1 Month
- 1 Month
- 1 Month
- 1 Month
- 1 Month
- 1 Month
- 1 Month
- 1 Month
- 1 Month
- 1 Month
- 1 Month
- 3 Months
- 3 Months
- 3 Months
- 1 Month
- 1 Month
- 1 Month
- 1 Month
- 1 Month
- 1 Month
- 1 Month
- 1 Month
- 3 Months
Specifications
Product Family Options
Product Information
- Connection TypeFitting materialFlow patternAvailabilityPrice
- MFLX31514-01Fitting Kit, HDPE, Straight/Reducer/Plug, Hose Barb, Small FittingsFitting Kit, HDPE, Straight/Reducer/Plug, Hose Barb, Small FittingsKitHDPEStraightLog in for availability1 * 1 ST122,00 €Specifications:CE certificate
More
Specifications
Cat. No.MFLX31514-01Model31514-01Max. temperature87 °C (190 °F)Min. temperature–53 °C (–65 °F)Warranty1 MonthCE certificate - MFLX31514-02Fitting Kit, Black Nylon, Straight/Reducer/Plug, Hosebarb, Small FittingsFitting Kit, Black Nylon, Straight/Reducer/Plug, Hosebarb, Small FittingsKitBlack NylonStraightLog in for availability1 * 1 ST137,00 €Specifications:
More
Specifications
Cat. No.MFLX31514-02Model31514-02Max. temperature93 °C (200 °F)Min. temperature–40 °C (–40 °F)Warranty1 Month - MFLX31514-05Fitting Kit, HDPE, Straight and Reducer, Hose Barb, Large FittingsFitting Kit, HDPE, Straight and Reducer, Hose Barb, Large FittingsKitHDPEStraightLog in for availability1 * 1 ST131,00 €Specifications:CE certificate
More
Specifications
Cat. No.MFLX31514-05Model31514-05Max. temperature87 °C (190 °F)Min. temperature–53 °C (–65 °F)Warranty1 MonthCE certificate - MFLX31514-06Fitting Kit, Black Nylon, Straight and Reducer, Hose Barb, Large Fittings; 1 SetFitting Kit, Black Nylon, Straight and Reducer, Hose Barb, Large Fittings; 1 SetKitNylonStraightLog in for availability1 * 1 ST126,00 €Specifications:
More
Specifications
Cat. No.MFLX31514-06Model31514-06Max. temperature93 °C (200 °F)Min. temperature–40 °C (–40 °F)Warranty1 Month - MFLX31514-09Fitting Kit, HDPE, Straight and Reducer, Threaded Adapter to Hose Barb; 1 SetFitting Kit, HDPE, Straight and Reducer, Threaded Adapter to Hose Barb; 1 SetKitHDPEStraightLog in for availability1 * 1 ST147,00 €Specifications:CE certificate
More
Specifications
Cat. No.MFLX31514-09Model31514-09Max. temperature87 °C (190 °F)Min. temperature–53 °C (–65 °F)Warranty1 MonthCE certificate - MFLX31514-10Fitting Kit, Black Nylon, Straight and Reducer, Thread Adapter to HosebarbFitting Kit, Black Nylon, Straight and Reducer, Thread Adapter to HosebarbKitNylonStraightLog in for availability1 * 1 ST146,00 €Specifications:
More
Specifications
Cat. No.MFLX31514-10Model31514-10Max. temperature93 °C (200 °F)Min. temperature–40 °C (–40 °F)Warranty1 Month - MFLX31514-13Fitting Kit, HDPE, Elbow/Tee/Y, Hose Barb; 1 SetFitting Kit, HDPE, Elbow/Tee/Y, Hose Barb; 1 SetKitHDPEElbowLog in for availability1 * 1 ST176,00 €Specifications:CE certificate
More
Specifications
Cat. No.MFLX31514-13Model31514-13Max. temperature87 °C (190 °F)Min. temperature–53 °C (–65 °F)Warranty1 MonthCE certificate - MFLX31514-14Fitting Kit, Black Nylon, Elbow/Tee/Y, Hose Barb; 1 SetFitting Kit, Black Nylon, Elbow/Tee/Y, Hose Barb; 1 SetKitNylonElbowLog in for availability1 * 1 ST141,00 €Specifications:
More
Specifications
Cat. No.MFLX31514-14Model31514-14Max. temperature93 °C (200 °F)Min. temperature–40 °C (–40 °F)Warranty1 Month - MFLX31514-17Fitting Kit, HDPE, Elbow and Tee, Hose Barb; 1 SetSpecifications:CE certificate
More
Specifications
Cat. No.MFLX31514-17Model31514-17Max. temperature87 °C (190 °F)Min. temperature–53 °C (–65 °F)Warranty1 MonthCE certificate - MFLX31514-18Fitting Kit, Black Nylon, Elbow and Tee, Hose Barb; 1 SetFitting Kit, Black Nylon, Elbow and Tee, Hose Barb; 1 SetKitNylonTLog in for availability1 * 1 ST120,00 €Specifications:
More
Specifications
Cat. No.MFLX31514-18Model31514-18Max. temperature93 °C (200 °F)Min. temperature–40 °C (–40 °F)Warranty1 Month - MFLX31514-22Fitting Kit, Black Nylon, Elbow/Tee/Nipple/Plug, Thread Adapter to HosebarbFitting Kit, Black Nylon, Elbow/Tee/Nipple/Plug, Thread Adapter to HosebarbKitBlack NylonElbowLog in for availability1 * 1 ST130,00 €Specifications:
More
Specifications
Cat. No.MFLX31514-22Model31514-22Max. temperature93 °C (200 °F)Min. temperature–40 °C (–40 °F)Warranty1 Month - MFLX31514-25Fitting Kit, HDPE, Elbow and Tee, Threaded Adapter to Hose Barb; 1 SetFitting Kit, HDPE, Elbow and Tee, Threaded Adapter to Hose Barb; 1 SetKitHDPETLog in for availability1 * 1 ST173,00 €Specifications:CE certificate
More
Specifications
Cat. No.MFLX31514-25Model31514-25Max. temperature87 °C (190 °F)Min. temperature–53 °C (–65 °F)Warranty1 MonthCE certificate - MFLX31514-26Fitting Kit, Black Nylon, Elbow and Tee, Threaded Adapter to Hose Barb; 1 SetFitting Kit, Black Nylon, Elbow and Tee, Threaded Adapter to Hose Barb; 1 SetKitBlack NylonTLog in for availability1 * 1 ST147,00 €Specifications:
More
Specifications
Cat. No.MFLX31514-26Model31514-26Max. temperature93 °C (200 °F)Min. temperature–40 °C (–40 °F)Warranty1 Month - MFLX31514-29Fitting Kit, Black Nylon, Straight/Reducer/Elbow/Tee, BSP Thread to HosebarbFitting Kit, Black Nylon, Straight/Reducer/Elbow/Tee, BSP Thread to HosebarbKitBlack NylonTLog in for availability1 * 1 ST146,00 €Specifications:
More
Specifications
Cat. No.MFLX31514-29Model31514-29Max. temperature93 °C (200 °F)Min. temperature–40 °C (–40 °F)Warranty1 Month - MFLX31514-32Fitting Kit, HDPE, Straight/Reducer/Elbow/Tee, BSP Threaded Adapter to HosebarbFitting Kit, HDPE, Straight/Reducer/Elbow/Tee, BSP Threaded Adapter to HosebarbKitHDPETLog in for availability1 * 1 ST773,00 €Specifications:CE certificate
More
Specifications
Cat. No.MFLX31514-32Model31514-32Max. temperature87 °C (190 °F)Min. temperature–53 °C (–65 °F)Warranty3 MonthsCE certificate - MFLX31514-33Fitting Kit, HDPE, Straight and Reducer, Threaded Adapter to Hose Barb and Threaded Adapter Union; 1 SetFitting Kit, HDPE, Straight and Reducer, Threaded Adapter to Hose Barb and Threaded Adapter Union; 1 SetKitHDPEStraightLog in for availability1 * 1 ST156,00 €Specifications:CE certificate
More
Specifications
Cat. No.MFLX31514-33Model31514-33Max. temperature87 °C (190 °F)Min. temperature–53 °C (–65 °F)Warranty3 MonthsCE certificate - MFLX31514-34Fitting Kit, Black Nylon, Straight and Reducer, Threaded Adapter to Hose Barb and Threaded Adapter Union; 1 SetFitting Kit, Black Nylon, Straight and Reducer, Threaded Adapter to Hose Barb and Threaded Adapter Union; 1 SetKitBlack NylonStraightLog in for availability1 * 1 ST158,00 €Specifications:
More
Specifications
Cat. No.MFLX31514-34Model31514-34Max. temperature93 °C (200 °F)Min. temperature–40 °C (–40 °F)Warranty3 Months - MFLX41514-03Fitting Kit, Polypropylene, Straight/Reducer/Plug, Hosebarb, Small FittingsFitting Kit, Polypropylene, Straight/Reducer/Plug, Hosebarb, Small FittingsKitPolypropyleneStraightLog in for availability1 * 1 ST138,00 €Specifications:CE certificate
More
Specifications
Cat. No.MFLX41514-03Model41514-03Max. temperature135 °C (275 °F)Min. temperature–17 °C (0 °F)Warranty1 MonthCE certificate - MFLX41514-07Fitting Kit, Polypropylene, Straight and Reducer, Hosebarb, Large FittingsFitting Kit, Polypropylene, Straight and Reducer, Hosebarb, Large FittingsKitPolypropyleneStraightLog in for availability1 * 1 ST138,00 €Specifications:CE certificate
More
Specifications
Cat. No.MFLX41514-07Model41514-07Max. temperature135 °C (275 °F)Min. temperature–17 °C (0 °F)Warranty1 MonthCE certificate - MFLX41514-11Fitting Kit, Polypropylene, Straight and Reducer, Threaded Adapter to HosebarbFitting Kit, Polypropylene, Straight and Reducer, Threaded Adapter to HosebarbKitPolypropyleneStraightLog in for availability1 * 1 ST160,00 €Specifications:CE certificate
More
Specifications
Cat. No.MFLX41514-11Model41514-11Max. temperature135 °C (275 °F)Min. temperature–17 °C (0 °F)Warranty1 MonthCE certificate - MFLX41514-15Fitting Kit, Polypropylene, Elbow/Tee/Y, Hose Barb; 1 SetFitting Kit, Polypropylene, Elbow/Tee/Y, Hose Barb; 1 SetKitPolypropyleneElbowLog in for availability1 * 1 ST178,00 €Specifications:CE certificate
More
Specifications
Cat. No.MFLX41514-15Model41514-15Max. temperature135 °C (275 °F)Min. temperature–17 °C (0 °F)Warranty1 MonthCE certificate - MFLX41514-19Fitting Kit, Polypropylene, Elbow and Tee, Hose Barb; 1 SetFitting Kit, Polypropylene, Elbow and Tee, Hose Barb; 1 SetKitPolypropyleneTLog in for availability1 * 1 ST164,00 €Specifications:CE certificate
More
Specifications
Cat. No.MFLX41514-19Model41514-19Max. temperature135 °C (275 °F)Min. temperature–17 °C (0 °F)Warranty1 MonthCE certificate - MFLX41514-23Fitting Kit, Polypropylene, Elbow/Tee/Nipple/Plug, Threaded Adapter to HosebarbFitting Kit, Polypropylene, Elbow/Tee/Nipple/Plug, Threaded Adapter to HosebarbKitPolypropyleneElbowLog in for availability1 * 1 ST160,00 €Specifications:CE certificate
More
Specifications
Cat. No.MFLX41514-23Model41514-23Max. temperature135 °C (275 °F)Min. temperature–17 °C (0 °F)Warranty1 MonthCE certificate - MFLX41514-27Fitting Kit, Polypropylene, Elbow and Tee, Threaded Adapter to Hose Barb; 1 SetFitting Kit, Polypropylene, Elbow and Tee, Threaded Adapter to Hose Barb; 1 SetKitPolypropyleneTLog in for availability1 * 1 ST173,00 €Specifications:CE certificate
More
Specifications
Cat. No.MFLX41514-27Model41514-27Max. temperature135 °C (275 °F)Min. temperature–17 °C (0 °F)Warranty1 MonthCE certificate - MFLX41514-30Fitting Kit, Polypropylene, Straight/Reducer/Elbow/Tee, BSP Thread to HosebarbFitting Kit, Polypropylene, Straight/Reducer/Elbow/Tee, BSP Thread to HosebarbKitPolypropyleneTLog in for availability1 * 1 ST164,00 €Specifications:CE certificate
More
Specifications
Cat. No.MFLX41514-30Model41514-30Max. temperature135 °C (275 °F)Min. temperature–17 °C (0 °F)Warranty1 MonthCE certificate - MFLX41514-35Fitting Kit, Polypropylene, Straight and Reducer, Threaded Adapter to Hose Barb and Threaded Adapter Union; 1 SetFitting Kit, Polypropylene, Straight and Reducer, Threaded Adapter to Hose Barb and Threaded Adapter Union; 1 SetKitPolypropyleneStraightLog in for availability1 * 1 ST176,00 €Specifications:CE certificate
More
Specifications
Cat. No.MFLX41514-35Model41514-35Max. temperature135 °C (275 °F)Min. temperature–17 °C (0 °F)Warranty3 MonthsCE certificate