ContactUsLinkComponent
Masterflex® Adapter Fittings, Hose Barb to Female NPT Threaded, Straight, Avantor®
Masterflex® Adapter Fittings, Hose Barb to Female NPT Threaded, Straight, Avantor®
Certificates
undefined
Masterflex® Adapter Fittings, Hose Barb to Female NPT Threaded, Straight, Avantor®
Specifications
  • Model:
  • 31501-40
  • 31500-41
  • 31701-85
  • 31500-40
  • 31702-22
  • 31702-20
  • 31501-41
  • Orifice dimensions:
  • 1,588 mm (0,0625")
  • 1,588 mm (0,0625")
  • 1,588 mm (0,0625")
  • 1,588 mm (0,0625")
  • Sterilisation:
  • Autoclave or ethylene oxide (ETO)
  • Ethylene oxide (ETO) only
  • Ethylene oxide (ETO) only
  • Ethylene oxide (ETO) only
  • Ethylene oxide (ETO) only
  • Ethylene oxide (ETO) only
  • Autoclave or ethylene oxide (ETO)
  • Warranty:
  • 1 year
  • 1 year
  • 1 year