Order Entry
Austria
ContactUsLinkComponent
MEM amino acids 50X solution without L-glutamine, Corning®
MEM amino acids 50X solution without L-glutamine, Corning®
Catalog # 25-030-CI
Supplier:  Corning
CAS Number:  
undefined
MEM amino acids 50X solution without L-glutamine, Corning®
Catalog # 25-030-CI
Supplier:  Corning
CAS Number:  

Specifications

  • Pk:
    6
  • Description:
    MEM Amino Acids 50X Solution without L-glutamine
  • Storage temperature:
    Frozen
  • Shipping temperature:
    Frozen

Specifications

About this item

The addition of MEM nonessential amino acids to medium can improve growth and the viability of propagating cells in culture while reducing the biosynthetic burden on these cells.

This concentrated supplement allows for efficient addition to media with no risk of dilution of media components.