1536-Well cyclic olefin copolymer microplates, Corning®
: Corning
1536-Well Cyclic Olefin Copolymer microplates are perfectly suited for high-throughput screening (HTS).
- Bar coded on the short side of the microplate
- Cyclic Olefin Copolymer (COC) material
- Flat top for improved sealing
- Ultra clear 127 µm film thickness
- Variety of treatment options
- Kalypsys and GNF system compatible
The ultraclear, 127 μm film bottom thickness is ideal for cell-based assays that require advanced optical analysis. The COC microplates are mechanically stable and inherently resistant to a broad range of solvents including DMSO and alcohols, allowing for compound storage as well.
A variety of surface treatments enable both cell-based and biochemical assays.
- Catalog No:
- CORN4677
- CORN4676
- CORN4675
- CORN4373
- CORN4372
- CORN4370
- CORN4371
- CORN4705
- Environmentally Preferable:
- Umweltschonende Herstellung
- Umweltschonende Herstellung
- Umweltzertifizierung:
- RECs, ISO 14001 - Kennebunk
- RECs, ISO 14001 - Kennebunk
Frequently Bought Together