PipeCheck standards for checking volumes of pipettes by photometry, Spectroquant®, Supelco®
Artikelnr. 1.14962.0001
Supplier: Merck
CAS Number:
Specifications
- Pk:25 Tests
- Description:Check solutions (4x6 cells) and reference solutions (4 cells ) for checking the volume of pipettes Spectroquant®
- M-Clarity quality level:MQ100