ESD safety shoes, OWEN, low, S2
Supplier: ELTEN
Certificates
About this item
Excellent wearing comfort, with an ankle-wrapping, softly padded upper edge that provides stability and grip.
- Hydrophobised microfibre/mesh material
- Fabric lining for natural ventilation
- Padded tongue
- Full-length inlay sole ESD PRO black
- ESD soft-fleece insole with integrated omissions
- TPU/PU sole WELLMAXX
- Composite toe cap
Excellent discharge capability, the shoe is suitable for work in ESD sensitive or electrostatically protected areas (EPA). Sole core made of Infinergy® by BASF. Leather-free.
Specifications
- Catalog No:
- ELTN729470/46
- ELTN729470/45
- ELTN729470/48
- ELTN729470/47
- ELTN729470/41
- ELTN729470/40
- ELTN729470/43
- ELTN729470/42
- ELTN729470/44
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001