ESD safety shoes, BRAYDEN XXTM, strap closure, low, S3
Supplier: ELTEN
Certificates
About this item
Excellent wearer comfort, with a padded upper edge to protect the Achilles’ tendon.
- Hydrophobised microfibre
- Fabric lining for natural ventilation
- Full-length inlay sole ESD PRO grey
- Non metallic, penetration resistant midsole
- MONO-PU sole WELLMAXX TRAINERS MONO
- Composite toe cap
Excellent discharge capability, the shoe is suitable for work in ESD-sensitive or electrostatically protected areas (EPA). Sole core made of Infinergy® by BASF. Metal- and leather-free.
Specifications
- Catalog No:
- 114-9130
- 114-9129
- 114-9131
- 114-9133
- 114-9132
- 114-9135
- 114-9134
- 114-9137
- 114-9136
- 114-9128
- 114-9139
- 114-9127
- 114-9138
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001