ESD safety ankle boots, TINE XXTL GTX, Mid, S2 CI
: ELTEN
This shoe is ideally tailored to the ergonomics of female feet. Excellent wearing comfort, with a padded upper edge to protect the Achilles tendon.
- Hydrophobised microfibre/Hydrophobized textile material
- GORE-TEX Performance Comfort Footwear
- Full, padded bellows tongue
- Full-length inlay sole ESD PRO Lady
- Soft fleece ESD insole
- Non-metallic, PU/PU sole WELLMAXX Trainers Lady
- Penetration-resistant midsole
- Steel toe cap and Achilles tendon protector
Thanks to its excellent discharge capability, the shoe is suitable for work in ESD sensitive or electrostatically protected areas (EPA). Sole core made of Infinergy® by BASF. Leather-free.
- Catalog No:
- ELTN746112/41
- ELTN746112/40
- ELTN746112/42
- ELTN746112/36
- ELTN746112/35
- ELTN746112/38
- ELTN746112/37
- ELTN746112/39
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
Frequently Bought Together