ESD safety ankle boots, DIALUTION, Mid, S3
Supplier: ELTEN
Certificates
About this item
Excellent wearing comfort, with a padded upper edge to protect the Achilles tendon.
- Hydrophobised microfibre
- Fabric lining for natural ventilation
- Full, padded bellows tongue
- Full-length inlay sole ESD PRO grey
- Non-metallic, penetration-resistant midsole
- MONO-PU sole WELLMAXX TRAINERS MONO
- Composite toe cap
Thanks to its excellent discharge capability, the shoe is suitable for work in ESD sensitive or electrostatically protected areas (EPA). Sole core made of Infinergy® by BASF. Leather-free.
Specifications
- Catalog No:
- ELTN760511/43
- ELTN760511/42
- ELTN760511/45
- ELTN760511/44
- ELTN760511/36
- ELTN760511/46
- ELTN760511/37
- ELTN760511/48
- ELTN760511/47
- ELTN760511/39
- ELTN760511/38
- ELTN760511/41
- ELTN760511/40
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001