Order Entry
Austria
ContactUsLinkComponent
Non return valve, PP
Non return valve, PP
Catalog # BURK8540-0012
Supplier:  BURKLE
Non return valve, PP
Catalog # BURK8540-0012
Supplier:  BURKLE

Specifications

  • Description:
    Non return valve, PP/POM, 1/2" thread
  • Pk:
    1

Specifications

About this item

Valve made of PP and POM, seal made of EPDM, springs made of stainless steel 1.4401.

  • Max. operating pressure 6 bar (at +20 °C)
  • Max. operating temperature 80 °C
  • Pressure difference 12 mbar