Order Entry
Austria
ContactUsLinkComponent
 
Defined fetal bovine Serum (FBS), HyClone™, U.S. origin
 
undefined
Defined fetal bovine Serum (FBS), HyClone™, U.S. origin

 

  • Catalog No:
  • SH30070.03IH30
  • SH30070.03IH25
  • SH30070.03IR25
  • SH30070.03IR30
  • Origin:
  • Animal
  • Animal
  • Animal
  • Animal
  • Endotoxin:
  • ≤10 EU/ml
  • ≤10 EU/ml
  • ≤10 EU/ml
  • ≤10 EU/ml
  • Haemoglobin:
  • ≤10 mg/dl
  • ≤10 mg/dl
  • ≤10 mg/dl
  • ≤10 mg/dl
  • Storage temperature:
  • 2...8 °C
  • 2...8 °C
  • 2...8 °C
  • 2...8 °C

 

Frequently Bought Together