Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Austria
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Penicillin/Streptomycin
Penicillin : Streptomycin solution (5000 IU/mL, 5 mg/mL), sterile
Penicillin : Streptomycin solution (5000 IU/mL, 5 mg/mL), sterile
Penicillin : Streptomycin solution (5000 IU/mL, 5 mg/mL), sterile
ICNA091670049
:
MP Biomedicals
:
Penicillin : Streptomycin solution (5000 IU/mL, 5 mg/mL), sterile
ICNA091670049
:
MP Biomedicals
:
Pk:
100 mL
Seq:
2440
:
:
GHS08
:
GHS07
:
Freezer
:
113-98-4 (Penicillin G potassium salt) - 5490-27-7 (Dihydrostreptomycin sulfate)
:
5490-27-7