ContactUsLinkComponent
Safety pipetting-ball
Safety pipetting-ball
Catalog # BURK7554-0002
Supplier:  BURKLE
undefined
Safety pipetting-ball
Catalog # BURK7554-0002
Supplier:  BURKLE
Specifications
  • Capacity:
    100 ml
  • Colour:
    Red
  • Environmentally Preferable:
  • Pk:
    1
Specifications
About this item
Peleus ball, simple pipetting aid. Evacuation takes place via an automatic valve. By pressing the valves, the liquid is drawn up, dispensed or purged.
  • For all pipettes up to 100 ml
The removable automatic valve also enables effective cleaning of the inside.