Sampling bags, SteriBag Cleanroom
Supplier: BURKLE
Certificates
About this item
The sampling bag SteriBag Cleanroom with double packaging can be placed directly in the cleanroom. 25 sample bags are placed in double secondary packaging. They can be removed one after the other and are therefore ideally suited for introducing into the cleanroom.
- Sterile, with double packaging
- RNase, DNase and pyrogen-free
- PE, transparent
- Liquid-tight
- With labelling field
Then the sterile bags can be used directly in the clean room. SteriBag Cleanroom is sterile inside and outside. This guarantees contamination-free storage and transport of solid, semi-solid and liquid media. The sample bag is perfect for use in chemical and pharmaceutical applications. The disposable sample bag has a labelling field, saves space and is also easy to transport. The large bag opening makes it easy to pour in the sample.
Frequently Bought Together