Order Entry
Austria
ContactUsLinkComponent
Sodium hydroxide 0.01 mol/l (0.01 N) in aqueous solution, AVS TITRINORM volumetric solution
Sodium hydroxide 0.01 mol/l (0.01 N) in aqueous solution, AVS TITRINORM volumetric solution
Catalog # 31631.290
Supplier:  VWR Chemicals
CAS Number:  
undefined
Sodium hydroxide 0.01 mol/l (0.01 N) in aqueous solution, AVS TITRINORM volumetric solution
Catalog # 31631.290
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    1 L
  • Seq:
    5037
  • Environmentally Preferable:

Specifications

Specification Test Results

  • Titer (20°C; real value 0.2 % accuracy):
    0.03992 - 0.04008 mol/l
  • NIST traceable:
    Confirmed

Specification Test Results

About this item

Synonyms: Caustic soda
 :
 :40 g/mol
Storage Temperature:Ambient
EINECS:215-185-5
CAS Number:1310-73-2
Formula:NaOH
MDL Number:MFCD00003548