ContactUsLinkComponent
Tamper-evident closure for compact jerrycan
Tamper-evident closure for compact jerrycan
Catalog # 215-4530
Supplier:  BURKLE
undefined
Tamper-evident closure for compact jerrycan
Catalog # 215-4530
Supplier:  BURKLE
Specifications
  • Thread:
    DIN 50
  • Colour:
    Blue
  • Description:
    Tamper-evident closure for compact jerrycan
  • Material:
    PP/PE
  • Pk:
    1
Specifications
About this item
Screw-on closure with tamper-evident ring for compact jerrycan.
  • Screw-on cover made of PP
  • Seal made out of PE
  • Thread DIN 50
The closure offers protection against tampering and a guarantee that the container has not been opened before, since the snap ring is visibly broken the first time the container is opened.