Order Entry
Austria
Orders LinkContactUsLinkComponent
Ethidium bromide solution 0,07 %

Ethidium bromide solution 0,07 %

Supplier:   PanReac AppliChem
Ethidium bromide solution 0,07 %
Supplier:  PanReac AppliChem
Certificates
Ethidium bromide solution 0,07 %

2 Options Available Below

Product Details & Documents

Ethidium bromide solutions with a concentration of 0,7 mg/ml.

Delivery information: Supplied in a dropper bottle.

Synonyms:  3,8-Diamino-5-ethyl-6-phenylphenanthridinium bromide, 3,8-Diamino-5-ethyl-6-phenylphenanthridin-5-ium bromide
  • CAS Number:1239-45-8
  • Formula:C₂₁H₂₀BrN₃
  • MDL Number:MFCD00011724
  • Molecular Weight:394.31 g/mol
  • Storage Temperature:Ambient
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Dropping bottle
    5 mL
    Log in for availability
    Log in for availability
    1 * 5 mL
    1 * 5 mL - 38,80 €
    38,80 €
    1 * 5 mL
       38,80 €

  • Dropping bottle
    15 mL
    Log in for availability
    Log in for availability
    1 * 15 mL
    1 * 15 mL - 77,20 €
    77,20 €
    1 * 15 mL
       77,20 €

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.