Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Austria
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Lids
Lids for Laboratory Vessels/Containers
Lids for buckets, square, HDPE
Lids for buckets, square, HDPE
Lids for buckets, square, HDPE
Catalog # SEMA0691
Supplier:
SEMADENI
Expand image
Lids for buckets, square, HDPE
Catalog # SEMA0691
Supplier:
SEMADENI
Specifications
Colour:
Black
Description:
Lid for bucket
L×W:
290×290 mm
Pk:
1
Specifications
About this item
Product Details
Product Details