PharmaFocus® Premium Silicone Tubing
Designed to meet a wide range of performance requirements in pharmaceutical applications. PharmaFocus® Premium Silicone Tubing is ideal for demanding fluid transfer processes and laboratory use. These products have been specifically formulated for use with sensitive media found in pharmaceutical and biotech processing.
- Performs in high purity fluid transfer and pump applications
- Tube resilience and adaptability due to excellent flexibility
- High visibility of fluid transport
- Versatile processing with cut-to-length, spools and packaging
- Biocompatibility for demanding biomedical / pharmaceutical applications
Typical hardness: 50 Shore
: Material certificate and lot traceability are included with every product. Full Compliance Statements, available upon request:
> Extractables test results - BPOG & USP 665
> USP Class VI / > ISO 10993
> European Pharmacopoeia 3.1.9
> USP 85, 87, 88, 661, 788 (ASTC, ASTP)
> Extractables test results - BPOG & USP 665
> USP Class VI / > ISO 10993
> European Pharmacopoeia 3.1.9
> USP 85, 87, 88, 661, 788 (ASTC, ASTP)
Frequently Bought Together