Orders LinkContactUsLinkComponent
Filling Solution, Reference, 1 M KNO₃
Filling Solution, Reference, 1 M KNO₃
Filling solution for reference electrode, 59 mL
Catalog # HACH28564026
Supplier:   Hach
Filling Solution, Reference, 1 M KNO₃
Catalog # HACH28564026
Supplier:  Hach
CAS Number:  
Filling Solution, Reference, 1 M KNO₃
Filling solution for reference electrode, 59 mL
CAS Number:  
Price
27,30 €
Log in for availability
1 * 59 mL
Product Details & Documents
Filling solution for electrode reference element, potassium nitrate (KNO₃), 1 M solution.
Specifications
Specifications
  • Pk:
    59 mL
  • Description:
    Filling solution for reference electrode