ContactUsLinkComponent
Industrial working at height safety helmet, EVOLite® Skyworker™
Industrial working at height safety helmet, EVOLite® Skyworker™
Catalog # JSPAAJS260-000-100
Supplier:  JSP
undefined
Industrial working at height safety helmet, EVOLite® Skyworker™
Catalog # JSPAAJS260-000-100
Supplier:  JSP
Specifications
  • Description:
    EVOLite® Skyworker™ Industrial working at height safety helmet
  • Pk:
    1
Specifications
About this item
The EVOLite® Skyworker™ helmet has a suspension system and shell structure that is designed for use in industrial, mountaineering, rescue, and leisure activities.
  • Industrial working at height safety helmet
  • Meets the EN 12492 mountaineering standard
  • Strong lightweight ABS shell
  • EPS liner for side, front, and rear impact protection
  • Fully adjustable 4-point chinstrap with quick release buckle
The EVOLite® Skyworker™ helmet conforms to the mandatory shock absorption and penetration requirements of EN 397 industrial standard. The helmet features a fully adjustable 4-point chinstrap with quick release buckle. Side, front and rear impact protection are provided by a shock-absorbing EPS liner. The liner is vented to increase comfort whilst maintaining a high protection factor.
The helmet also features a 6-point Terylene harness system and Chamlon™ sweatband. Revolution® wheel ratchet, 3-D adjustment, and a vented shell reduce heat within the helmet.
Certifications: EN 397, EN 12492.