Orders LinkContactUsLinkComponent
Elasticated chinstrap for JSP helmets
Elasticated chinstrap for JSP helmets
Elasticated chinstrap
Artikelnr. JSPAAHV001-100-000
Anbieter:   JSP
Elasticated chinstrap for JSP helmets
Artikelnr. JSPAAHV001-100-000
Anbieter:  JSP
Elasticated chinstrap for JSP helmets
Elasticated chinstrap for JSP helmets
Elasticated chinstrap
Preis
30,90 €
Log in for availability
1 * 10 ST
Product Details & Documents
Elasticated chinstrap releases automatically with forces between 150 and 250 Newtons.
  • Wide elastic chinstrap
  • Releases automatically with forces between 150 and 250 Newtons
  • Nylon and elastic webbing
  • HDPE clips
Fits all Evolution® safety helmets.
Certifications: EN 397, EN 812.
Specifications
  • Description:
    Elasticated chinstrap