Clear safety spectacles, Crackerjack™
Crackerjack™ safety spectacles, Clear
Catalog # JSPA1CRAK23C
Supplier: JSP
Jump to:
Product Details & Documents
Stylish, single lens, lightweight safety spectacle.
- Ultra enveloping
- Lateral protection
- Flat temples
The Crackerjack™ is a stylish single lens spectacle that is lightweight with flat temples providing comfort for extended periods of use. Its design makes it a perfect fit for wearing in conjunction with helmets and ear defenders.
Certifications: Meets EN 166:2001.T9:V9
Documents
TDS Spectacles Crackerjack
Datasheet
Specifications
- Description:Crackerjack™ safety spectacles
- Seq:01
- Lenses:Clear