Order Entry
Austria
ContactUsLinkComponent
Recombinant Human Glutathione Peroxidase 1
Recombinant Human Glutathione Peroxidase 1
Supplier:  SELENOZYME
Certificates
undefined
Recombinant Human Glutathione Peroxidase 1
Supplier:  SELENOZYME

Specifications

  • Storage conditions:
    Protein stored in 25 mM Tris-HCl (pH 8.0) with 250 mM KCl, 2 mM 2-Mercaptoethanol and 10% glycerol
  • Purity:
    ≥ 95%
  • Concentration:
    Concentration 1 mg protein/ml buffer.

Specifications

Related Information

Frequently Bought Together