Sampling head, MD8 Airscan®
MD8 air samplers stainless steel sampling Head Bayonet, stainless steel with silicone seal
Artikelnr. 514-1293
Anbieter: Sartorius
Jump to:
Product Details & Documents
Sampling head with bayonet connection.
- Flexible operating conditions
- Generates a vacuum and pumps a defined air sample volume from the ambient air
The bayonet connector accommodates the filter adapter. Sampling heads with a bayonet connection are particularly suitable for applications where the sampling head is not installed in a system.
Ordering information: Supplied without accessories.
Specifications
- Description:MD8 air samplers stainless steel sampling Head Bayonet, stainless steel with silicone seal