Orders LinkContactUsLinkComponent
Writable Cryo Colour Rectangles and Dots
Supplier:   GA
Writable Cryo Colour Rectangles and Dots
Supplier:  GA
Certificates
Writable Cryo Colour Rectangles and Dots
Writable Cryo Colour Rectangles and Dots
61 Options Available Below
Product Details & Documents
Writable cryogenic colour dots and rectangles provided in dispenser box rolls for identifying the sides and tops of cryo vials and microtubes stored in lab freezers and liquid nitrogen dewars. Easy to write on with permanent cryo markers, JTRA-class labels are available in a variety of colours.
  • Temperature range: −196 to +110 °C (−320.8 to +230 °F)
  • Designed for use in liquid nitrogen (−196 °C), freezers (−80 °C), and dry ice
  • Ideal for eppendorf microtubes and cryo vials
  • Strong adhesive ensures labels won't wrinkle or peel during long-term storage
  • Available in a wide range of colours, for color-coding and easy identification
These writable cryogenic rectangle and dot labels for cryo vials and conical bottom microtubes are made with a strong permanent adhesive ideal for liquid nitrogen, lab freezers, and dry ice storage and transportation. Easily writable with permanent cryogenic markers, these ultra-low temperature labels are supplied in roll format and are available in 15 individual colours as well as packs of 15 assorted colours for colour coding and rapid identification.
Product Family Options
Circle permanent cryogenic writable labels
Product Information
  • Colour
    Dimensions
    Packed
    Availability
    Price
  • GAINJTRA-9BE
    Cryogenic writable label, core size 1"
    Cryogenic writable label, core size 1"
    Beige
    9 mm (0,35")
    1100 labels/roll (2 labels across)
    Log in for availability
    Log in for availability
    1 * 1 Rolle
    1 * 1 Rolle - 35,20 €
    35,20 €
    1 * 1 Rolle
       35,20 €
  • GAINJTRA-9BL
    Cryogenic writable label, core size 1"
    Cryogenic writable label, core size 1"
    Blue
    9 mm (0,35")
    1100 labels/roll (2 labels across)
    Log in for availability
    Log in for availability
    1 * 1 Rolle
    1 * 1 Rolle - 35,20 €
    35,20 €
    1 * 1 Rolle
       35,20 €
  • GAINJTRA-9BR
    Cryogenic writable label, core size 1"
    Cryogenic writable label, core size 1"
    Brown
    9 mm (0,35")
    1100 labels/roll (2 labels across)
    Log in for availability
    Log in for availability
    1 * 1 Rolle
    1 * 1 Rolle - 35,20 €
    35,20 €
    1 * 1 Rolle
       35,20 €
  • GAINJTRA-9GA
    Cryogenic writable label, core size 1"
    Cryogenic writable label, core size 1"
    Green
    9 mm (0,35")
    1100 labels/roll (2 labels across)
    Log in for availability
    Log in for availability
    1 * 1 Rolle
    1 * 1 Rolle - 35,20 €
    35,20 €
    1 * 1 Rolle
       35,20 €
  • GAINJTRA-9GS
    Cryogenic writable label, core size 1"
    Cryogenic writable label, core size 1"
    Light green
    9 mm (0,35")
    1100 labels/roll (2 labels across)
    Log in for availability
    Log in for availability
    1 * 1 Rolle
    1 * 1 Rolle - 35,20 €
    35,20 €
    1 * 1 Rolle
       35,20 €
  • GAINJTRA-9LA
    Cryogenic writable label, core size 1"
    Cryogenic writable label, core size 1"
    Light purple
    9 mm (0,35")
    1100 labels/roll (2 labels across)
    Log in for availability
    Log in for availability
    1 * 1 Rolle
    1 * 1 Rolle - 35,20 €
    35,20 €
    1 * 1 Rolle
       35,20 €
  • GAINJTRA-9LE
    Cryogenic writable label, core size 1"
    Cryogenic writable label, core size 1"
    Light yellow
    9 mm (0,35")
    1100 labels/roll (2 labels across)
    Log in for availability
    Log in for availability
    1 * 1 Rolle
    1 * 1 Rolle - 35,20 €
    35,20 €
    1 * 1 Rolle
       35,20 €
  • GAINJTRA-9MT
    Cryogenic writable label, core size 1"
    Cryogenic writable label, core size 1"
    Mint green
    9 mm (0,35")
    1100 labels/roll (2 labels across)
    Log in for availability
    Log in for availability
    1 * 1 Rolle
    1 * 1 Rolle - 35,20 €
    35,20 €
    1 * 1 Rolle
       35,20 €
  • GAINJTRA-9OR
    Cryogenic writable label, core size 1"
    Cryogenic writable label, core size 1"
    Orange
    9 mm (0,35")
    1100 labels/roll (2 labels across)
    Log in for availability
    Log in for availability
    1 * 1 Rolle
    1 * 1 Rolle - 35,20 €
    35,20 €
    1 * 1 Rolle
       35,20 €
  • GAINJTRA-9PI
    Cryogenic writable label, core size 1"
    Cryogenic writable label, core size 1"
    Pink
    9 mm (0,35")
    1100 labels/roll (2 labels across)
    Log in for availability
    Log in for availability
    1 * 1 Rolle
    1 * 1 Rolle - 35,20 €
    35,20 €
    1 * 1 Rolle
       35,20 €
  • GAINJTRA-9RE
    Cryogenic writable label, core size 1"
    Cryogenic writable label, core size 1"
    Red
    9 mm (0,35")
    1100 labels/roll (2 labels across)
    Log in for availability
    Log in for availability
    1 * 1 Rolle
    1 * 1 Rolle - 35,20 €
    35,20 €
    1 * 1 Rolle
       35,20 €
  • GAINJTRA-9SI
    Cryogenic writable label, core size 1"
    Cryogenic writable label, core size 1"
    Silver
    9 mm (0,35")
    1100 labels/roll (2 labels across)
    Log in for availability
    Log in for availability
    1 * 1 Rolle
    1 * 1 Rolle - 35,20 €
    35,20 €
    1 * 1 Rolle
       35,20 €
  • GAINJTRA-9VI
    Cryogenic writable label, core size 1"
    Cryogenic writable label, core size 1"
    Violet
    9 mm (0,35")
    1100 labels/roll (2 labels across)
    Log in for availability
    Log in for availability
    1 * 1 Rolle
    1 * 1 Rolle - 35,20 €
    35,20 €
    1 * 1 Rolle
       35,20 €
  • GAINJTRA-9WH
    Cryogenic writable label, core size 1"
    Cryogenic writable label, core size 1"
    White
    9 mm (0,35")
    1100 labels/roll (2 labels across)
    Log in for availability
    Log in for availability
    1 * 1 Rolle
    1 * 1 Rolle - 35,20 €
    35,20 €
    1 * 1 Rolle
       35,20 €
  • GAINJTRA-9YE
    Cryogenic writable label, core size 1"
    Cryogenic writable label, core size 1"
    Yellow
    9 mm (0,35")
    1100 labels/roll (2 labels across)
    Log in for availability
    Log in for availability
    1 * 1 Rolle
    1 * 1 Rolle - 35,20 €
    35,20 €
    1 * 1 Rolle
       35,20 €
  • GAINJTRA-11BE
    Cryogenic writable label, core size 1"
    Cryogenic writable label, core size 1"
    Beige
    11 mm (0,44")
    1100 labels/roll (2 labels across)
    Log in for availability
    Log in for availability
    1 * 420 Rolle
    1 * 420 Rolle - 35,20 €
    35,20 €
    1 * 420 Rolle
       35,20 €
  • GAINJTRA-11BL
    Cryogenic writable label, core size 1"
    Cryogenic writable label, core size 1"
    Blue
    11 mm (0,44")
    1100 labels/roll (2 labels across)
    Log in for availability
    Log in for availability
    1 * 1 Rolle
    1 * 1 Rolle - 35,20 €
    35,20 €
    1 * 1 Rolle
       35,20 €
  • GAINJTRA-11BR
    Cryogenic writable label, core size 1"
    Cryogenic writable label, core size 1"
    Brown
    11 mm (0,44")
    1100 labels/roll (2 labels across)
    Log in for availability
    Log in for availability
    1 * 1 Rolle
    1 * 1 Rolle - 35,20 €
    35,20 €
    1 * 1 Rolle
       35,20 €
  • GAINJTRA-11GA
    Cryogenic writable label, core size 1"
    Cryogenic writable label, core size 1"
    Green
    11 mm (0,44")
    1100 labels/roll (2 labels across)
    Log in for availability
    Log in for availability
    1 * 1 Rolle
    1 * 1 Rolle - 35,20 €
    35,20 €
    1 * 1 Rolle
       35,20 €
  • GAINJTRA-11GS
    Cryogenic writable label, core size 1"
    Cryogenic writable label, core size 1"
    Light green
    11 mm (0,44")
    1100 labels/roll (2 labels across)
    Log in for availability
    Log in for availability
    1 * 1 Rolle
    1 * 1 Rolle - 35,20 €
    35,20 €
    1 * 1 Rolle
       35,20 €
  • GAINJTRA-11LA
    Cryogenic writable label, core size 1"
    Cryogenic writable label, core size 1"
    Light purple
    11 mm (0,44")
    1100 labels/roll (2 labels across)
    Log in for availability
    Log in for availability
    1 * 1 Rolle
    1 * 1 Rolle - 35,20 €
    35,20 €
    1 * 1 Rolle
       35,20 €
  • GAINJTRA-11LE
    Cryogenic writable label, core size 1"
    Cryogenic writable label, core size 1"
    Light yellow
    11 mm (0,44")
    1100 labels/roll (2 labels across)
    Log in for availability
    Log in for availability
    1 * 1 Rolle
    1 * 1 Rolle - 35,20 €
    35,20 €
    1 * 1 Rolle
       35,20 €
  • GAINJTRA-11MT
    Cryogenic writable label, core size 1"
    Cryogenic writable label, core size 1"
    Mint green
    11 mm (0,44")
    1100 labels/roll (2 labels across)
    Log in for availability
    Log in for availability
    1 * 1 Rolle
    1 * 1 Rolle - 35,20 €
    35,20 €
    1 * 1 Rolle
       35,20 €
  • GAINJTRA-11OR
    Cryogenic writable label, core size 1"
    Cryogenic writable label, core size 1"
    Orange
    11 mm (0,44")
    1100 labels/roll (2 labels across)
    Log in for availability
    Log in for availability
    1 * 1 Rolle
    1 * 1 Rolle - 35,20 €
    35,20 €
    1 * 1 Rolle
       35,20 €
  • GAINJTRA-11PI
    Cryogenic writable label, core size 1"
    Cryogenic writable label, core size 1"
    Pink
    11 mm (0,44")
    1100 labels/roll (2 labels across)
    Log in for availability
    Log in for availability
    1 * 1 Rolle
    1 * 1 Rolle - 35,20 €
    35,20 €
    1 * 1 Rolle
       35,20 €
  • GAINJTRA-11RE
    Cryogenic writable label, core size 1"
    Cryogenic writable label, core size 1"
    Red
    11 mm (0,44")
    1100 labels/roll (2 labels across)
    Log in for availability
    Log in for availability
    1 * 1 Rolle
    1 * 1 Rolle - 35,20 €
    35,20 €
    1 * 1 Rolle
       35,20 €
  • GAINJTRA-11SI
    Cryogenic writable label, core size 1"
    Cryogenic writable label, core size 1"
    Silver
    11 mm (0,44")
    1100 labels/roll (2 labels across)
    Log in for availability
    Log in for availability
    1 * 1 Rolle
    1 * 1 Rolle - 35,20 €
    35,20 €
    1 * 1 Rolle
       35,20 €
  • GAINJTRA-11VI
    Cryogenic writable label, core size 1"
    Cryogenic writable label, core size 1"
    Violet
    11 mm (0,44")
    1100 labels/roll (2 labels across)
    Log in for availability
    Log in for availability
    1 * 1 Rolle
    1 * 1 Rolle - 35,20 €
    35,20 €
    1 * 1 Rolle
       35,20 €
  • GAINJTRA-11WH
    Cryogenic writable label, core size 1"
    Cryogenic writable label, core size 1"
    White
    11 mm (0,44")
    1100 labels/roll (2 labels across)
    Log in for availability
    Log in for availability
    1 * 1 Rolle
    1 * 1 Rolle - 35,20 €
    35,20 €
    1 * 1 Rolle
       35,20 €
  • GAINJTRA-11YE
    Cryogenic writable label, core size 1"
    Cryogenic writable label, core size 1"
    Yellow
    11 mm (0,44")
    1100 labels/roll (2 labels across)
    Log in for availability
    Log in for availability
    1 * 1 Rolle
    1 * 1 Rolle - 35,20 €
    35,20 €
    1 * 1 Rolle
       35,20 €
  • GAINJTRA-13BE
    Cryogenic writable label, core size 1"
    Cryogenic writable label, core size 1"
    Beige
    12,7 mm (0,5")
    1100 labels/roll (2 labels across)
    Log in for availability
    Log in for availability
    1 * 1 Rolle
    1 * 1 Rolle - 35,20 €
    35,20 €
    1 * 1 Rolle
       35,20 €
  • GAINJTRA-13BL
    Cryogenic writable label, core size 1"
    Cryogenic writable label, core size 1"
    Blue
    12,7 mm (0,5")
    1100 labels/roll (2 labels across)
    Log in for availability
    Log in for availability
    1 * 1 Rolle
    1 * 1 Rolle - 35,20 €
    35,20 €
    1 * 1 Rolle
       35,20 €
  • GAINJTRA-13BR
    Cryogenic writable label, core size 1"
    Cryogenic writable label, core size 1"
    Brown
    12,7 mm (0,5")
    1100 labels/roll (2 labels across)
    Log in for availability
    Log in for availability
    1 * 1 Rolle
    1 * 1 Rolle - 35,20 €
    35,20 €
    1 * 1 Rolle
       35,20 €
  • GAINJTRA-13GA
    Cryogenic writable label, core size 1"
    Cryogenic writable label, core size 1"
    Green
    12,7 mm (0,5")
    1100 labels/roll (2 labels across)
    Log in for availability
    Log in for availability
    1 * 1 Rolle
    1 * 1 Rolle - 35,20 €
    35,20 €
    1 * 1 Rolle
       35,20 €
  • GAINJTRA-13GS
    Cryogenic writable label, core size 1"
    Cryogenic writable label, core size 1"
    Light green
    12,7 mm (0,5")
    1100 labels/roll (2 labels across)
    Log in for availability
    Log in for availability
    1 * 1 Rolle
    1 * 1 Rolle - 35,20 €
    35,20 €
    1 * 1 Rolle
       35,20 €
  • GAINJTRA-13LA
    Cryogenic writable label, core size 1"
    Cryogenic writable label, core size 1"
    Light purple
    12,7 mm (0,5")
    1100 labels/roll (2 labels across)
    Log in for availability
    Log in for availability
    1 * 1 Rolle
    1 * 1 Rolle - 35,20 €
    35,20 €
    1 * 1 Rolle
       35,20 €
  • GAINJTRA-13LE
    Cryogenic writable label, core size 1"
    Cryogenic writable label, core size 1"
    Light yellow
    12,7 mm (0,5")
    1100 labels/roll (2 labels across)
    Log in for availability
    Log in for availability
    1 * 1 Rolle
    1 * 1 Rolle - 35,20 €
    35,20 €
    1 * 1 Rolle
       35,20 €
  • GAINJTRA-13MT
    Cryogenic writable label, core size 1"
    Cryogenic writable label, core size 1"
    Mint green
    12,7 mm (0,5")
    1100 labels/roll (2 labels across)
    Log in for availability
    Log in for availability
    1 * 1 Rolle
    1 * 1 Rolle - 35,20 €
    35,20 €
    1 * 1 Rolle
       35,20 €
  • GAINJTRA-13OR
    Cryogenic writable label, core size 1"
    Cryogenic writable label, core size 1"
    Orange
    12,7 mm (0,5")
    1100 labels/roll (2 labels across)
    Log in for availability
    Log in for availability
    1 * 1 Rolle
    1 * 1 Rolle - 35,20 €
    35,20 €
    1 * 1 Rolle
       35,20 €
  • GAINJTRA-13PI
    Cryogenic writable label, core size 1"
    Cryogenic writable label, core size 1"
    Pink
    12,7 mm (0,5")
    1100 labels/roll (2 labels across)
    Log in for availability
    Log in for availability
    1 * 1 Rolle
    1 * 1 Rolle - 35,20 €
    35,20 €
    1 * 1 Rolle
       35,20 €
  • GAINJTRA-13RE
    Cryogenic writable label, core size 1"
    Cryogenic writable label, core size 1"
    Red
    12,7 mm (0,5")
    1100 labels/roll (2 labels across)
    Log in for availability
    Log in for availability
    1 * 1 Rolle
    1 * 1 Rolle - 35,20 €
    35,20 €
    1 * 1 Rolle
       35,20 €
  • GAINJTRA-13SI
    Cryogenic writable label, core size 1"
    Cryogenic writable label, core size 1"
    Silver
    12,7 mm (0,5")
    1100 labels/roll (2 labels across)
    Log in for availability
    Log in for availability
    1 * 1 Rolle
    1 * 1 Rolle - 35,20 €
    35,20 €
    1 * 1 Rolle
       35,20 €
  • GAINJTRA-13VI
    Cryogenic writable label, core size 1"
    Cryogenic writable label, core size 1"
    Violet
    12,7 mm (0,5")
    1100 labels/roll (2 labels across)
    Log in for availability
    Log in for availability
    1 * 1 Rolle
    1 * 1 Rolle - 35,20 €
    35,20 €
    1 * 1 Rolle
       35,20 €
  • GAINJTRA-13WH
    Cryogenic writable label, core size 1"
    Cryogenic writable label, core size 1"
    White
    12,7 mm (0,5")
    1100 labels/roll (2 labels across)
    Log in for availability
    Log in for availability
    1 * 1 Rolle
    1 * 1 Rolle - 35,20 €
    35,20 €
    1 * 1 Rolle
       35,20 €
  • GAINJTRA-13X6A
    Cryogenic writable label, core size 1"
    Cryogenic writable label, core size 1"
    Multicolour
    12,7 mm (0,5")
    5040 labels/roll (6 labels across)
    Log in for availability
    Log in for availability
    1 * 1 Rolle
    1 * 1 Rolle - 144,00 €
    144,00 €
    1 * 1 Rolle
       144,00 €
  • GAINJTRA-13YE
    Cryogenic writable label, core size 1"
    Cryogenic writable label, core size 1"
    Yellow
    12,7 mm (0,5")
    1100 labels/roll (2 labels across)
    Log in for availability
    Log in for availability
    1 * 1 Rolle
    1 * 1 Rolle - 35,20 €
    35,20 €
    1 * 1 Rolle
       35,20 €
Rectangle permanent cryogenic writable labels
Product Information
  • Colour
    Dimensions
    Packed
    Availability
    Price
  • GAINJTRA-2610BE
    Cryogenic writable label, core size 1"
    Cryogenic writable label, core size 1"
    Beige
    26×10 mm (1,02×0,4")
    1000 labels/roll (1 label across)
    Log in for availability
    Log in for availability
    1 * 1 Rolle
    1 * 1 Rolle - 35,20 €
    35,20 €
    1 * 1 Rolle
       35,20 €
  • GAINJTRA-2610BL
    Cryogenic writable label, core size 1"
    Cryogenic writable label, core size 1"
    Blue
    26×10 mm (1,02×0,4")
    1000 labels/roll (1 label across)
    Log in for availability
    Log in for availability
    1 * 1 Rolle
    1 * 1 Rolle - 35,20 €
    35,20 €
    1 * 1 Rolle
       35,20 €
  • GAINJTRA-2610BR
    Cryogenic writable label, core size 1"
    Cryogenic writable label, core size 1"
    Brown
    26×10 mm (1,02×0,4")
    1000 labels/roll (1 label across)
    Log in for availability
    Log in for availability
    1 * 1 Rolle
    1 * 1 Rolle - 35,20 €
    35,20 €
    1 * 1 Rolle
       35,20 €
  • GAINJTRA-2610GA
    Cryogenic writable label, core size 1"
    Cryogenic writable label, core size 1"
    Green
    26×10 mm (1,02×0,4")
    1000 labels/roll (1 label across)
    Log in for availability
    Log in for availability
    1 * 1 Rolle
    1 * 1 Rolle - 35,20 €
    35,20 €
    1 * 1 Rolle
       35,20 €
  • GAINJTRA-2610GS
    Cryogenic writable label, core size 1"
    Cryogenic writable label, core size 1"
    Light green
    26×10 mm (1,02×0,4")
    1000 labels/roll (1 label across)
    Log in for availability
    Log in for availability
    1 * 1 Rolle
    1 * 1 Rolle - 35,20 €
    35,20 €
    1 * 1 Rolle
       35,20 €
  • GAINJTRA-2610LA
    Cryogenic writable label, core size 1"
    Cryogenic writable label, core size 1"
    Light purple
    26×10 mm (1,02×0,4")
    1000 labels/roll (1 label across)
    Log in for availability
    Log in for availability
    1 * 1 Rolle
    1 * 1 Rolle - 35,20 €
    35,20 €
    1 * 1 Rolle
       35,20 €
  • GAINJTRA-2610LE
    Cryogenic writable label, core size 1"
    Cryogenic writable label, core size 1"
    Light yellow
    26×10 mm (1,02×0,4")
    1000 labels/roll (1 label across)
    Log in for availability
    Log in for availability
    1 * 1 Rolle
    1 * 1 Rolle - 35,20 €
    35,20 €
    1 * 1 Rolle
       35,20 €
  • GAINJTRA-2610MT
    Cryogenic writable label, core size 1"
    Cryogenic writable label, core size 1"
    Mint green
    26×10 mm (1,02×0,4")
    1000 labels/roll (1 label across)
    Log in for availability
    Log in for availability
    1 * 1 Rolle
    1 * 1 Rolle - 35,20 €
    35,20 €
    1 * 1 Rolle
       35,20 €
  • GAINJTRA-2610OR
    Cryogenic writable label, core size 1"
    Cryogenic writable label, core size 1"
    Orange
    26×10 mm (1,02×0,4")
    1000 labels/roll (1 label across)
    Log in for availability
    Log in for availability
    1 * 1 Rolle
    1 * 1 Rolle - 35,20 €
    35,20 €
    1 * 1 Rolle
       35,20 €
  • GAINJTRA-2610PI
    Cryogenic writable label, core size 1"
    Cryogenic writable label, core size 1"
    Pink
    26×10 mm (1,02×0,4")
    1000 labels/roll (1 label across)
    Log in for availability
    Log in for availability
    1 * 1 Rolle
    1 * 1 Rolle - 35,20 €
    35,20 €
    1 * 1 Rolle
       35,20 €
  • GAINJTRA-2610RE
    Cryogenic writable label, core size 1"
    Cryogenic writable label, core size 1"
    Red
    26×10 mm (1,02×0,4")
    1000 labels/roll (1 label across)
    Log in for availability
    Log in for availability
    1 * 1 Rolle
    1 * 1 Rolle - 35,20 €
    35,20 €
    1 * 1 Rolle
       35,20 €
  • GAINJTRA-2610SI
    Cryogenic writable label, core size 1"
    Cryogenic writable label, core size 1"
    Silver
    26×10 mm (1,02×0,4")
    1000 labels/roll (1 label across)
    Log in for availability
    Log in for availability
    1 * 1 Rolle
    1 * 1 Rolle - 35,20 €
    35,20 €
    1 * 1 Rolle
       35,20 €
  • GAINJTRA-2610VI
    Cryogenic writable label, core size 1"
    Cryogenic writable label, core size 1"
    Violet
    26×10 mm (1,02×0,4")
    1000 labels/roll (1 label across)
    Log in for availability
    Log in for availability
    1 * 1 Rolle
    1 * 1 Rolle - 35,20 €
    35,20 €
    1 * 1 Rolle
       35,20 €
  • GAINJTRA-2610WH
    Cryogenic writable label, core size 1"
    Cryogenic writable label, core size 1"
    White
    26×10 mm (1,02×0,4")
    1000 labels/roll (1 label across)
    Log in for availability
    Log in for availability
    1 * 1 Rolle
    1 * 1 Rolle - 35,20 €
    35,20 €
    1 * 1 Rolle
       35,20 €
  • GAINJTRA-2610YE
    Cryogenic writable label, core size 1"
    Cryogenic writable label, core size 1"
    Yellow
    26×10 mm (1,02×0,4")
    1000 labels/roll (1 label across)
    Log in for availability
    Log in for availability
    1 * 1 Rolle
    1 * 1 Rolle - 35,20 €
    35,20 €
    1 * 1 Rolle
       35,20 €