Orders LinkContactUsLinkComponent
DYMO-compatible cryogenic labels
DYMO-compatible cryogenic labels
Supplier:   GA
DYMO-compatible cryogenic labels
Supplier:  GA
Certificates
DYMO-compatible cryogenic labels
20 Options Available Below
Product Details & Documents
DYMO compatible, direct thermal cryo labels. For ultra-low and cryogenic temperature applications as low as –196 °C.
  • EG1F-081 series labels are resistant to gamma irradiation (tested to up to 50 kGy)
  • Ideal for the identification of 0,5 to 5 ml vials and tubes
  • Compatible with all DYMO label writers
  • Temperature range: –196 to 70 °C
  • Permanent adhesive
Ideal for identification and permanent storage of vials and containers in liquid nitrogen and laboratory freezers (–80, –50, –20 °C). Labels will also resist extreme cryogenic temperatures (–196 °C in a liquid phase and vapor phase liquid nitrogen).
Product Family Options
Product Information
  • Colour
    Dimensions
    Availability
    Price
  • GAINEG1F-072BE
    Rectangle cryo labels
    Rectangle cryo labels
    Beige
    12,7×26×9,5 mm
    Log in for availability
    Log in for availability
    1 * 1 Rolle
    1 * 1 Rolle - 99,00 €
    99,00 €
    1 * 1 Rolle
       99,00 €
  • GAINEG1F-072BL
    Rectangle cryo labels
    Rectangle cryo labels
    Blue
    12,7×26×9,5 mm
    Log in for availability
    Log in for availability
    1 * 1 Rolle
    1 * 1 Rolle - 99,00 €
    99,00 €
    1 * 1 Rolle
       99,00 €
  • GAINEG1F-072BR
    Cryo labels
    Cryo labels
    Brown
    12,7×26×9,5 mm
    Log in for availability
    Log in for availability
    1 * 1 Rolle
    1 * 1 Rolle - 99,00 €
    99,00 €
    1 * 1 Rolle
       99,00 €
  • GAINEG1F-072GA
    Cryo labels
    Cryo labels
    Green
    12,7×26×9,5 mm
    Log in for availability
    Log in for availability
    1 * 1 Rolle
    1 * 1 Rolle - 99,00 €
    99,00 €
    1 * 1 Rolle
       99,00 €
  • GAINEG1F-072GR
    Cryo labels
    Cryo labels
    Green
    12,7×26×9,5 mm
    Log in for availability
    Log in for availability
    1 * 1 Rolle
    1 * 1 Rolle - 99,00 €
    99,00 €
    1 * 1 Rolle
       99,00 €
  • GAINEG1F-072LE
    Cryo labels
    Cryo labels
    Yellow
    12,7×26×9,5 mm
    Log in for availability
    Log in for availability
    1 * 1 Rolle
    1 * 1 Rolle - 99,00 €
    99,00 €
    1 * 1 Rolle
       99,00 €
  • GAINEG1F-072OR
    Rectangle cryo labels
    Rectangle cryo labels
    Orange
    12,7×26×9,5 mm
    Log in for availability
    Log in for availability
    1 * 1 Rolle
    1 * 1 Rolle - 99,00 €
    99,00 €
    1 * 1 Rolle
       99,00 €
  • GAINEG1F-072RE
    Rectangle cryo labels
    Rectangle cryo labels
    Red
    12,7×26×9,5 mm
    Log in for availability
    Log in for availability
    1 * 1 Rolle
    1 * 1 Rolle - 99,00 €
    99,00 €
    1 * 1 Rolle
       99,00 €
  • GAINEG1F-072VI
    Rectangle cryo labels
    Rectangle cryo labels
    Violet
    12,7×26×9,5 mm
    Log in for availability
    Log in for availability
    1 * 1 Rolle
    1 * 1 Rolle - 99,00 €
    99,00 €
    1 * 1 Rolle
       99,00 €
  • GAINEG1F-072YE
    Rectangle cryo labels
    Rectangle cryo labels
    Yellow
    12,7×26×9,5 mm
    Log in for availability
    Log in for availability
    1 * 1 Rolle
    1 * 1 Rolle - 99,00 €
    99,00 €
    1 * 1 Rolle
       99,00 €
  • GAINEG1F-081BE
    Round cryo labels
    Round cryo labels
    Beige
    26,4×12,7×9,5 mm circle
    Log in for availability
    Log in for availability
    1 * 1 Rolle
    1 * 1 Rolle - 99,00 €
    99,00 €
    1 * 1 Rolle
       99,00 €
  • GAINEG1F-081BL
    Round cryo labels
    Round cryo labels
    Blue
    26,4×12,7×9,5 mm circle
    Log in for availability
    Log in for availability
    1 * 1 Rolle
    1 * 1 Rolle - 99,00 €
    99,00 €
    1 * 1 Rolle
       99,00 €
  • GAINEG1F-081BR
    Round cryo labels
    Round cryo labels
    Brown
    26,4×12,7×9,5 mm circle
    Log in for availability
    Log in for availability
    1 * 1 Rolle
    1 * 1 Rolle - 99,00 €
    99,00 €
    1 * 1 Rolle
       99,00 €
  • GAINEG1F-081GA
    Rectangle labels plus round cryo tags
    Rectangle labels plus round cryo tags
    Green
    26,4×12,7×9,5 mm circle
    Log in for availability
    Log in for availability
    1 * 1 Rolle
    1 * 1 Rolle - 99,00 €
    99,00 €
    1 * 1 Rolle
       99,00 €
  • GAINEG1F-081GR
    Rectangle labels plus round cryo tags
    Rectangle labels plus round cryo tags
    Green
    26,4×12,7×9,5 mm circle
    Log in for availability
    Log in for availability
    1 * 1 Rolle
    1 * 1 Rolle - 99,00 €
    99,00 €
    1 * 1 Rolle
       99,00 €
  • GAINEG1F-081LE
    Rectangle labels plus round cryo tags
    Rectangle labels plus round cryo tags
    Yellow
    26,4×12,7×9,5 mm circle
    Log in for availability
    Log in for availability
    1 * 1 Rolle
    1 * 1 Rolle - 99,00 €
    99,00 €
    1 * 1 Rolle
       99,00 €
  • GAINEG1F-081OR
    Rectangle labels plus round cryo tags
    Rectangle labels plus round cryo tags
    Orange
    26,4×12,7×9,5 mm circle
    Log in for availability
    Log in for availability
    1 * 1 Rolle
    1 * 1 Rolle - 99,00 €
    99,00 €
    1 * 1 Rolle
       99,00 €
  • GAINEG1F-081RE
    Round cryo labels
    Round cryo labels
    Red
    26,4×12,7×9,5 mm circle
    Log in for availability
    Log in for availability
    1 * 1 Rolle
    1 * 1 Rolle - 99,00 €
    99,00 €
    1 * 1 Rolle
       99,00 €
  • GAINEG1F-081VI
    Round cryo labels
    Round cryo labels
    Violet
    26,4×12,7×9,5 mm circle
    Log in for availability
    Log in for availability
    1 * 1 Rolle
    1 * 1 Rolle - 99,00 €
    99,00 €
    1 * 1 Rolle
       99,00 €
  • GAINEG1F-081YE
    Round cryo labels
    Round cryo labels
    Yellow
    26,4×12,7×9,5 mm circle
    Log in for availability
    Log in for availability
    1 * 1 Rolle
    1 * 1 Rolle - 99,00 €
    99,00 €
    1 * 1 Rolle
       99,00 €