Orders LinkContactUsLinkComponent
Face shield visor, 701.02
Face shield visor, 701.02
Interchangeable visor, Anti-fog, Acetate
Catalog # UNVT1220010
Supplier:   UNIVET
Face shield visor, 701.02
Catalog # UNVT1220010
Supplier:  UNIVET
Face shield visor, 701.02
Face shield visor, 701.02
Interchangeable visor, Anti-fog, Acetate
Price
104,00 €
Log in for availability
1 * 5 ST
Product Details & Documents
The 701.02 face shield meets rigorous EN 166 standards (droplets and splashes) by providing visor with extended coverage on the upper part of the forehead.
  • Anti-fog acetate visor
  • Adjustable elastic head band
  • Visor inclines up to 90°
  • Interchangeable visor
  • Face shield suitable for use in combination with prescription glasses and protective masks
This anti-fog visor allows maximum vision due to anti-fog characteristics. A soft sponge in front with an easily adjustable band ensures comfort. This visor is suitable for use with corrective glasses and respiratory protective masks.
Certifications: Certified according to EN 166, marking 3 protection against droplets and liquid splash.
Specifications
  • Type:
    Anti-fog
  • Description:
    Interchangeable visor
  • Material:
    Acetate