Orders LinkContactUsLinkComponent
Utility Boxes
Utility Boxes
Supplier:   MULTIROIR
Utility Boxes
Supplier:  MULTIROIR
Certificates
Utility Boxes
5 Options Available Below
Product Details & Documents
Range of practical PP boxes in 9 different sizes, 5 round and 4 square, ranging from 0,25 to 8 L.
  • Food grade
  • Soft and flexible lids
  • Anti-slip surface suitable for freezer and fridge
  • Acid and alkali resistant
  • Cold and heat resistant: –20 to +100 °C
All the boxes have a soft and flexible lid, ensuring a fastened closure as well as easy assembling and removal.
The surface of the boxes is slightly roughed and offers a 'anti-slip' function even with wet hands.
Easy to clean, these utility laboratory storage boxes are stackable and can be placed in the freezer or the fridge.
Product Family Options
Utility box, round
Product Information
  • Capacity
    W×D×H
    Availability
    Price
  • 0,5 L
    110×110×94 mm
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 38,80 €
    38,80 €
    1 * 10 ST
       38,80 €
  • 1 L
    134×134×112 mm
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 54,00 €
    54,00 €
    1 * 10 ST
       54,00 €
  • 2 L
    167×167×145 mm
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 85,50 €
    85,50 €
    1 * 10 ST
       85,50 €
  • 2 L
    103×103×171 mm
    Log in for availability
    Log in for availability
    1 * 6 ST
    1 * 6 ST - 45,30 €
    45,30 €
    1 * 6 ST
       45,30 €
Utility box, rectangular
Product Information
  • Capacity
    W×D×H
    Availability
    Price
  • 5 L
    283×283×95 mm
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - 70,60 €
    70,60 €
    1 * 5 ST
       70,60 €