ESD safety shoes, lace-up, S3, jo_Speedy Low, Jori
Supplier: ELTEN
Certificates
About this item
Excellent wearing comfort, and suitable for work in ESD sensitive or electrostatically protected areas.
- Padded tongue
- Non-metallic, penetration-resistant midsole
- Rubber/EVA sole jo_DYNAMIC
- Rubber scuff cap
- Composite toe cap
Hydrophobised textile material with fabric lining for natural ventilation. Full-length inlay ESD sole. Metal-free and leather-free.
Certifications: EN ISO 20345 S3 HRO/SRC, form A.
Specifications
- Catalog No:
- ELTN12151/39
- ELTN12151/40
- ELTN12151/41
- ELTN12151/42
- ELTN12151/36
- ELTN12151/37
- ELTN12151/38
- ELTN12161/36
- ELTN12161/37
- ELTN12161/48
- ELTN12151/43
- ELTN12151/44
- ELTN12151/45
- ELTN12161/38
- ELTN12161/39
- ELTN12151/46
- ELTN12151/47
- ELTN12161/40
- ELTN12161/43
- ELTN12161/41
- ELTN12151/48
- ELTN12161/42
- ELTN12161/44
- ELTN12161/45
- ELTN12161/46
- ELTN12161/47
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001