Nitrogen generator, Precision Nitrogen Headspace 250
564-0298
- Description:Precision Nitrogen Headspace 250 nitrogen generator
- Pk:1
The Precision Nitrogen Headspace 250 has been conceived and custom-engineered to meet the gas requirements for headspace sampler instruments requiring higher gas pressure.
- Ultra High Purity nitrogen for headspace sampler instruments
- Can supply make-up gas
- Single unit can stack with the Precision Hydrogen, Nitrogen and Zero Air generators
- Service and repair status indicators
- 1 year comprehensive on-site warranty