ContactUsLinkComponent
Nitrogen and dry air gas generator, Genius 3045
Nitrogen and dry air gas generator, Genius 3045
Catalog # 564-0280
Supplier:  PEAK SCIENTIFIC
undefined
Nitrogen and dry air gas generator, Genius 3045
Catalog # 564-0280
Supplier:  PEAK SCIENTIFIC
Specifications
  • Description:
    Genius 3045 nitrogen and dry air gas generator
  • Pk:
    1
Specifications
About this item
Designed to the specifications of the Bruker EVOQ triple Quadrupole's gas requirements delivering dry air at up to 50 L/min and nitrogen at up to 32 L/min.
  • Independent nitrogen and dry air gas supplies can be tailored for its application
  • Higher flow 30 series generator designed for hungrier LC-MS applications
  • Compressor based solution, no need for an external air supply
  • Highly economical source of nitrogen gas with low lifetime running costs
  • Gas is supplied on demand so generator works to your schedule
  • Service indication to allow planning of preventative maintenance
  • Minimal set-up required
  • The latest generation of compressors located in an insulated chamber reducing noise and vibration
  • 1 year comprehensive on-site warranty