ContactUsLinkComponent
Grade 2589 D filter papers for technical use, Whatman™
Grade 2589 D filter papers for technical use, Whatman™
Certificates
undefined
Grade 2589 D filter papers for technical use, Whatman™
Specifications
  • Catalog No:
  • WHAT10343978
  • 512-0411
  • Grade:
  • 2589 d
  • 2589 d
  • Weight:
  • 500 g/m²
  • 500 g/m²
  • Thickness:
  • 1000 µm
  • 1000 µm
  • Particle retention:
  • 2 - 6 µm
  • 2 - 6 µm
  • Filtration speed:
  • 235 s (Herzberg)
  • 235 s (Herzberg)