Orders LinkContactUsLinkComponent
Nitrile gloves, Kimtech™ Opal™
Nitrile gloves, Kimtech™ Opal™
   Sustainable Options Available
Supplier:   Ansell
Nitrile gloves, Kimtech™ Opal™
Supplier:  Ansell
   Sustainable Options Available
Certificates
Nitrile gloves, Kimtech™ Opal™
5 Options Available Below
Product Details & Documents
Kimtech™ Opal™ nitrile gloves guard against contamination by chemical splash and microorganism hazards. Powder-free, ambidextrous, with beaded cuff and textured fingertips. The absence of natural rubber latex and vulcanising agents reduces the risk of TYPE I and IV glove associated skin reactions.
  • Assured compliance
  • Formulated without thiurams, thiazoles, guanidines, carbamates, and no sulphur added
  • Soft, comfortable, and easy to don
  • Textured fingertips enhance grip and tactile sensitivity for safer and more efficient processes
  • Dark colour to enhance concentration on task
  • Available in a wide range of sizes, from XS to XL
These gloves guard against contamination by chemical splash and microorganism hazards, delivering seamless protection when and where it counts. Powder-free gloves that are ideal for use in research and production facilities, life sciences, and non sterile drug manufacturing applications. The gloves feature an innovative approach using a thin nitrile polymer with a tip thickness of just 0,07 mm, but with excellent tensile strength suitable for rigorous process use. Easy to don, soft and comfortable, they are suitable for extended wearing time.
The absence of natural rubber latex, sulphur and vulcanising agents reduces the risk of TYPE I and TYPE IV glove associated skin reactions, protecting the wearer as well as the application. Precision manufacturing reduces the risk of contamination and ensures the nitrile gloves offer high levels of regulatory compliance. Beaded cuffs and textured fingertips enable excellent handling of both wet and dry materials, and the ambidextrous gloves have been extensively tested under stringent conditions. In addition, the thin nitrile construction and efficient packaging mean that the chemical resistant gloves can reduce waste by up to 50%.
Certifications: EN ISO 374-1 Type B (KPT), EN 374-4, EN ISO 374-5 (VIRUS), Cat. III; AQL 1,5
Food contact approved
Documents
Specifications for Product Family
Specifications
  • Catalog No:
  • 112-6223
  • 112-6224
  • 112-6225
  • 112-6226
  • 112-6227
  • Environmentally Preferable:
  • Waste Reducing
  • Waste Reducing
  • Waste Reducing
  • Waste Reducing
  • Waste Reducing
  • Ambidextrous / hand-specific:
  • Ambidextrous
  • Ambidextrous
  • Ambidextrous
  • Ambidextrous
  • Ambidextrous
  • Disposable / reusable:
  • Disposable
  • Disposable
  • Disposable
  • Disposable
  • Disposable
  • Latex / latex-free:
  • Latex-free
  • Latex-free
  • Latex-free
  • Latex-free
  • Latex-free
  • Powdered / powder-free:
  • Powder-free
  • Powder-free
  • Powder-free
  • Powder-free
  • Powder-free
  • Material:
  • Nitrile
  • Nitrile
  • Nitrile
  • Nitrile
  • Nitrile
  • Finish:
  • Textured fingertips
  • Textured fingertips
  • Textured fingertips
  • Textured fingertips
  • Textured fingertips
  • Cuff style:
  • Beaded
  • Beaded
  • Beaded
  • Beaded
  • Beaded
  • EN norm:
  • EN ISO 374-1 Type B (KPT), EN 374-4, EN ISO 374-5 (VIRUS)
  • EN ISO 374-1 Type B (KPT), EN 374-4, EN ISO 374-5 (VIRUS)
  • EN ISO 374-1 Type B (KPT), EN 374-4, EN ISO 374-5 (VIRUS)
  • EN ISO 374-1 Type B (KPT), EN 374-4, EN ISO 374-5 (VIRUS)
  • EN ISO 374-1 Type B (KPT), EN 374-4, EN ISO 374-5 (VIRUS)
  • Food contact approved:
  • +
  • +
  • +
  • +
  • +
Specifications
Product Family Options
Product Information
  • Glove size
    Colour
    Length
    Packed
    Availability
    Price
  • XS
    Dark blue
    240 mm
    200/box
    Log in for availability
    Log in for availability
    1 * 200 ST
    1 * 200 ST - 37,70 €
    37,70 €
    1 * 200 ST
       37,70 €
    Specifications:
      Waste Reducing
    More
    Specifications
    Cat. No.
    112-6223
    Environmentally Preferable
    Waste Reducing
    Ambidextrous / hand-specific
    Ambidextrous
    Disposable / reusable
    Disposable
    Latex / latex-free
    Latex-free
    Powdered / powder-free
    Powder-free
    Material
    Nitrile
    Finish
    Textured fingertips
    Cuff style
    Beaded
    EN norm
    EN ISO 374-1 Type B (KPT), EN 374-4, EN ISO 374-5 (VIRUS)
    Food contact approved
    +
      Waste Reducing
  • S
    Dark blue
    240 mm
    200/box
    Log in for availability
    Log in for availability
    1 * 200 ST
    1 * 200 ST - 37,70 €
    37,70 €
    1 * 200 ST
       37,70 €
    Specifications:
      Waste Reducing
    More
    Specifications
    Cat. No.
    112-6224
    Environmentally Preferable
    Waste Reducing
    Ambidextrous / hand-specific
    Ambidextrous
    Disposable / reusable
    Disposable
    Latex / latex-free
    Latex-free
    Powdered / powder-free
    Powder-free
    Material
    Nitrile
    Finish
    Textured fingertips
    Cuff style
    Beaded
    EN norm
    EN ISO 374-1 Type B (KPT), EN 374-4, EN ISO 374-5 (VIRUS)
    Food contact approved
    +
      Waste Reducing
  • M
    Dark blue
    240 mm
    200/box
    Log in for availability
    Log in for availability
    1 * 200 ST
    1 * 200 ST - 37,70 €
    37,70 €
    1 * 200 ST
       37,70 €
    Specifications:
      Waste Reducing
    More
    Specifications
    Cat. No.
    112-6225
    Environmentally Preferable
    Waste Reducing
    Ambidextrous / hand-specific
    Ambidextrous
    Disposable / reusable
    Disposable
    Latex / latex-free
    Latex-free
    Powdered / powder-free
    Powder-free
    Material
    Nitrile
    Finish
    Textured fingertips
    Cuff style
    Beaded
    EN norm
    EN ISO 374-1 Type B (KPT), EN 374-4, EN ISO 374-5 (VIRUS)
    Food contact approved
    +
      Waste Reducing
  • L
    Dark blue
    240 mm
    200/box
    Log in for availability
    Log in for availability
    1 * 200 ST
    1 * 200 ST - 37,70 €
    37,70 €
    1 * 200 ST
       37,70 €
    Specifications:
      Waste Reducing
    More
    Specifications
    Cat. No.
    112-6226
    Environmentally Preferable
    Waste Reducing
    Ambidextrous / hand-specific
    Ambidextrous
    Disposable / reusable
    Disposable
    Latex / latex-free
    Latex-free
    Powdered / powder-free
    Powder-free
    Material
    Nitrile
    Finish
    Textured fingertips
    Cuff style
    Beaded
    EN norm
    EN ISO 374-1 Type B (KPT), EN 374-4, EN ISO 374-5 (VIRUS)
    Food contact approved
    +
      Waste Reducing
  • XL
    Dark blue
    240 mm
    170/box
    Log in for availability
    Log in for availability
    1 * 170 ST
    1 * 170 ST - 32,00 €
    32,00 €
    1 * 170 ST
       32,00 €
    Specifications:
      Waste Reducing
    More
    Specifications
    Cat. No.
    112-6227
    Environmentally Preferable
    Waste Reducing
    Ambidextrous / hand-specific
    Ambidextrous
    Disposable / reusable
    Disposable
    Latex / latex-free
    Latex-free
    Powdered / powder-free
    Powder-free
    Material
    Nitrile
    Finish
    Textured fingertips
    Cuff style
    Beaded
    EN norm
    EN ISO 374-1 Type B (KPT), EN 374-4, EN ISO 374-5 (VIRUS)
    Food contact approved
    +
      Waste Reducing