Orders LinkContactUsLinkComponent
XF Matrix, CELLvo™
XF Matrix, CELLvo™
Supplier:   STEMBIOSYS
XF Matrix, CELLvo™
Supplier:  STEMBIOSYS
Certificates
XF Matrix, CELLvo™
XF Matrix, CELLvo™
3 Options Available Below
Product Details & Documents
CELLvo™ XF Matrix is a ready to use cell culture substrate which provides a xeno-free native microenvironment that replicates the 3-dimensional 'home' in which stem cells naturally reside and proliferate.
  • Matrix made of polystyrene
  • Mesenchymal stem cells grown on CELLvo™ XF Matrix proliferate more rapidly
  • Mesenchymal stem cells grown on CELLvo™ XF Matrix maintain a smaller cell size
  • Mesenchymal stem cells grown on CELLvo™ XF Matrix exhibit a higher percentage of cells expressing SSEA-4
CELLvo™ XF Matrix is a natural extracellular matrix synthesised in vitro by bone marrow stromal cells and includes the biochemistry and architecture normally found in the bone marrow. CELLvo™ XF Matrix is a protein extracellular matrix produced under xeno-free conditions that mimics the normal microenvironment of stem cells. This 3-D matrix is produced by young adult human bone marrow cells. The final product is a thin, off-white film on cell culture flasks, dishes, and substrates.
Accessories information: Accessories not included.
Delivery information: CELLvo™ XF Matrix arrives at room temperature but should be refrigerated at 2 to 8 °C upon arrival. Avoid extended exposure to light.
Packaging: CELLvo™ XF Matrix is produced under aseptic conditions and should be handled using aseptic techniques.
Caution: CELLvo™ XF Matrix is not for human use. This product should not be used for diagnostic or therapeutic purposes. Handle CELLvo™ related products in accordance with established bio-safety practices.
Product Family Options
Product Information
  • Size
    Pk
    Availability
    Price
  • STBSXF-HPME-6WP
    6-well plate, flat bottom, cylindrical wells, non-pyrogenic, 86×128 mm (footprint), growth area 9,5 cm²
    6-well plate, flat bottom, cylindrical wells, non-pyrogenic, 86×128 mm (footprint), growth area 9,5 cm²
    15.5 ml (total volume/Well)
    5
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - 229,00 €
    229,00 €
    1 * 5 ST
       229,00 €
  • STBSXF-HPME-T75
    U-shaped culture flask, canted, vented, non-pyrogenic, growth area 75 cm²
    U-shaped culture flask, canted, vented, non-pyrogenic, growth area 75 cm²
    5
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - 229,00 €
    229,00 €
    1 * 5 ST
       229,00 €
  • STBSXF-HPME-T150
    Culture flask, canted, vented, non-pyrogenic, growth area 150 cm²
    Culture flask, canted, vented, non-pyrogenic, growth area 150 cm²
    30 - 45 ml (medium volume)
    5
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - 344,00 €
    344,00 €
    1 * 5 ST
       344,00 €