Order Entry
Austria
ContactUsLinkComponent
Matrix, CELLvo™
Matrix, CELLvo™
Supplier:  STEMBIOSYS
Certificates
undefined
Matrix, CELLvo™
Supplier:  STEMBIOSYS

Specifications

  • Source:
  • Species:
  • Environmentally Preferable:

Specifications

Frequently Bought Together