Accessories for Continuous Level Control, SymLine®
About this item
LED's and acoustic alarm warn you in time before the fill level of your collection containers reaches the critical level. With mute / snooze function and interface for controlling external devices. For installation in your media line of the fume cupboard.
- Built-in Signalbox2 for Safety Waste Cap and other electronical level controlers
- Touchless alarm-resetting fields for two containers
- Two switch inputs and two switch outputs for peripheral units
- Incl. 230 V power supply cable with open end and clamp
Suitable e.g. for equipment from Wesemann, Waldner, Bense, Hemling, Köttermann, Kewaunee, Carlo Erba, Modus Complete, Delagrave, and many more.
5 Options Available Below
Product Details & Documents
LED's and acoustic alarm warn you in time before the fill level of your collection containers reaches the critical level. With mute / snooze function and interface for controlling external devices. For installation in your media line of the fume cupboard.
- Built-in Signalbox2 for Safety Waste Cap and other electronical level controlers
- Touchless alarm-resetting fields for two containers
- Two switch inputs and two switch outputs for peripheral units
- Incl. 230 V power supply cable with open end and clamp
Suitable e.g. for equipment from Wesemann, Waldner, Bense, Hemling, Köttermann, Kewaunee, Carlo Erba, Modus Complete, Delagrave, and many more.
Documents
GHS Poster Important Lab and HPLC Safety Guidelines
Application Notes
Product Family Options
Product Information
- AvailabilityPrice
- SCTV106548Built-in Signalbox2; 2-channel (EU), for Safety waste cap and other electronic level controlers, touchless alarm-resetting fields for 2 containers, 2 switch inputs and 2 switch outputs for peripheral units incl. 230 V power supply cable with open end and clamp, universal plugBuilt-in Signalbox2; 2-channel (EU), for Safety waste cap and other electronic level controlers, touchless alarm-resetting fields for 2 containers, 2 switch inputs and 2 switch outputs for peripheral units incl. 230 V power supply cable with open end and clamp, universal plugLog in for availability1 * 1 ST456,75 €
- SCTV106698Built-in signal box table stand, brushed stainless steel, 2 mm wall thicknessBuilt-in signal box table stand, brushed stainless steel, 2 mm wall thicknessLog in for availability1 * 1 ST58,30 €
- SCTV106741Built-in Signalbox1, EU/UK/US, 1-channel, for safety waste cap and other electronical level controlers, touchless alarm-resetting fields for one container, one switch input and one switch output for peripheral units, including 24 V power supplyBuilt-in Signalbox1, EU/UK/US, 1-channel, for safety waste cap and other electronical level controlers, touchless alarm-resetting fields for one container, one switch input and one switch output for peripheral units, including 24 V power supplyLog in for availability1 * 1 ST304,50 €
- SCTV106733Built-in signal box display, brushed stainless steel, 2 mm wall thickness, for signal boxes 1/2/5 with cable connection on the back (Version 2024)Built-in signal box display, brushed stainless steel, 2 mm wall thickness, for signal boxes 1/2/5 with cable connection on the back (Version 2024)Log in for availability1 * 1 ST58,30 €
- SCTV106735Built-in Signalbox5, EU/UK/US, 5-channel, for safety waste cap and other electronical level controlers, touchless alarm-resetting fields for five containers, five switch inputs and five switch outputs for peripheral units, including 24 V power supplyBuilt-in Signalbox5, EU/UK/US, 5-channel, for safety waste cap and other electronical level controlers, touchless alarm-resetting fields for five containers, five switch inputs and five switch outputs for peripheral units, including 24 V power supplyLog in for availability1 * 1 ST578,55 €



