Collection containers, Corning®
Lieferant: Corning
Zertifikate
Über diesen Artikel
Corning® collection containers are fabricated to match specific application requirements, and are available in a variety of sizes and connector configurations.
- Single-use bags
- Material ULDPE
- Designed to be practical and cost-effective alternatives to rigid-walled containers
- Ideal for biopharmaceutical applications
Unless otherwise noted in container port descriptions, all port tubing components are chemically resistant, heat sealable flexible tubing.
Port 1: 60 L × ¹/₂ Int.Ø ×³/₄ Ext.Ø tubing, Male MPX connector, Female MPC cap, pinch clamp.
Port 2: 40 L × ¹/₈ Int.Ø ×¹/₄ Ext.Ø tubing, Female Luer with ⁵/₃₂" Barb, Swabable valve, Non-breather Female Luer cap.
Port 3: 60 L × ¹/₂ Int.Ø ×³/₄ Ext.Ø tubing, 1,5" Tri-clover with ¹/₂" Barb, pinch clamp
Spezifikationen
- Catalog No:
- MDTC91-200-20
- MDTC91-200-36
- MDTC91-200-10
- MDTC91-200-02
- MDTC91-200-48
- MDTC91-200-05
- MDTC91-200-82
- MDTC91-200-45
- MDTC91-200-01
- MDTC91-200-47
- MDTC91-200-83
- MDTC91-200-42
- MDTC91-100-30
- MDTC91-200-43
- MDTC91-200-39
- MDTC91-001-MB
- MDTC91-200-41
- Port:
- Port 1: Male Luer connector with ¹/₈ Barb + cap, 2' L × ¹/₈ Int.Ø ×¹/₄ Ext.Ø tubing, Slide clamp
Port 2: Male MPC connector with ³/₈ Barb + cap, 6 L × ³/₈ Int.Ø ×⁵/₈" Ext.Ø PCMGS tubing, Slide clamp
Port 3: Female Luer with ³/₁₆ Barb, Swabbable valve, cap - Port 1: Female MPC connector with ¹/₄ Barb + Plug, Slide clamp
Port 2: Male MPC connector with ¹/₄ Barb + cap, Slide clamp
Port 3: Vinyl cap, Swabbable valve, Female Luer with Barb - Port 1: Male Luer connector with ¹/₈ Barb + cap, 2' L × ¹/₈ Int.Ø ×¹/₄ Ext.Ø tubing, Slide clamp
Port 2: Male MPC connector with ³/₈ Barb + cap, 6 L × ³/₈ Int.Ø ×⁵/₈" Ext.Ø PCMGS tubing, Slide clamp
Port 3: Female Luer with ³/₁₆ Barb, Swabbable valve, cap - Port 1: Male Luer connector with ¹/₈ Barb + cap, 2' L × ¹/₈ Int.Ø ×¹/₄ Ext.Ø tubing, Slide clamp
Port 2: Male MPC connector with ³/₈ Barb + cap, 6 L × ³/₈ Int.Ø ×⁵/₈" Ext.Ø PCMGS tubing, Slide clamp
Port 3: Female Luer with ³/₁₆ Barb, Swabbable valve, cap - Port 1: ³/₈ connector, 12 L × ³/₈ Int.Ø ×⁵/₈" Ext.Ø tubing, Male MPC connector with ³/₈" Barb, Female MPC cap
Port 2: ³/₈ connector, 12 L × ³/₈ Int.Ø ×⁵/₈" Ext.Ø tubing, Female MPC connector with ³/₈" Barb, Male MPC plug
Port 3: Break-off injection site - Port 1: Male Luer connector with ¹/₈ Barb + cap, 2' L × ¹/₈ Int.Ø ×¹/₄ Ext.Ø tubing, Slide clamp
Port 2: Male MPC connector with ³/₈ Barb + cap, 6 L × ³/₈ Int.Ø ×⁵/₈" Ext.Ø PCMGS tubing, Slide clamp
Port 3: Female Luer with ³/₁₆ Barb, Swabbable valve, cap - Port 1: 60 L × ¹/₂ Int.Ø ×³/₄ Ext.Ø tubing, Male MPX connector, Female MPC cap, pinch clamp
Port 2: 40 L × ¹/₈ Int.Ø ×¹/₄ Ext.Ø tubing, Female Luer with ⁵/₃₂" Barb, Swabbable valve, Non-breather Female Luer cap
Port 3: 60 L × ¹/₂ Int.Ø ×³/₄ Ext.Ø tubing, 1.5" Tri-clover with ¹/₂" Barb, pinch clamp - Port 1: ³/₈ connector, 12 L × ³/₈ Int.Ø ×⁵/₈" Ext.Ø tubing, Male MPC connector with ³/₈" Barb, Female MPC cap
Port 2: ³/₈ connector, 12 L × ³/₈ Int.Ø ×⁵/₈" Ext.Ø tubing, Female MPC connector with ³/₈" Barb, Male MPC plug
Port 3: Break-off injection site - Port 1: Male Luer connector with ¹/₈ Barb + cap, 2' L × ¹/₈ Int.Ø ×¹/₄ Ext.Ø tubing, Slide clamp
Port 2: Male MPC connector with ³/₈ Barb + cap, 6 L × ³/₈ Int.Ø ×⁵/₈" Ext.Ø PCMGS tubing, Slide clamp
Port 3: Female Luer with ³/₁₆ Barb, Swabbable valve, cap - Port 1: ³/₈ connector, 12 L × ³/₈ Int.Ø ×⁵/₈" Ext.Ø tubing, Male MPC connector with ³/₈" Barb, Female MPC cap
Port 2: ³/₈ connector, 12 L × ³/₈ Int.Ø ×⁵/₈" Ext.Ø tubing, Female MPC connector with ³/₈" Barb, Male MPC plug
Port 3: Break-off injection site - Port 1: 60 L × ¹/₂ Int.Ø ×³/₄ Ext.Ø tubing, Male MPX connector, Female MPC cap, pinch clamp
Port 2: 40 L × ¹/₈ Int.Ø ×¹/₄ Ext.Ø tubing, Female Luer with ⁵/₃₂" Barb, Swabbable valve, Non-breather Female Luer cap
Port 3: 60 L × ¹/₂ Int.Ø ×³/₄ Ext.Ø tubing, 1.5" Tri-clover with ¹/₂" Barb, pinch clamp - Port 1: Break-off injection site
Port 2: Female MPC connector with ¹/₄ Barb, Male MPC plug
Port 3: Male MPC connector with ¹/₄ Barb, Female MPC cap - Port 1: 24 L × ³/₈ Int.Ø ×⁵/₈ Ext.Ø tubing, Male CPC connector with ³/₈" barb, PVC dust cover cap, pinch clamp
Port 2: 24 L × ³/₈ Int.Ø ×⁵/₈ Ext.Ø tubing, Male CPC connector with ³/₈" barb, PVC dust cover cap, pinch clamp
Port 3: 36 L × ¹/₈ Int.Ø ×¹/₄ Ext.Ø tubing, Female Luer with ¹/₈" Barb, Male Luer cap - Port 1: ³/₈ connector, 12 L × ³/₈ Int.Ø ×⁵/₈" Ext.Ø tubing, Male MPC connector with ³/₈" Barb, Female MPC cap
Port 2: ³/₈ connector, 12 L × ³/₈ Int.Ø ×⁵/₈" Ext.Ø tubing, Female MPC connector with ³/₈" Barb, Male MPC plug
Port 3: Break-off injection site - Port 1: Female MPC connector with ¹/₄ Barb + Plug, Slide clamp
Port 2: Male MPC connector with ¹/₄ Barb + cap, Slide clamp
Port 3: Vinyl cap, Swabbable valve, Female Luer with Barb - Port 1: 24 L × ¹/₄ Int.Ø ×³/₈" Ext.Ø PCMGS tubing, Female Luer with ¹/₄" barb, cap, slide clamp
Port 2: 24 L × ¹/₄ Int.Ø ×³/₈" Ext.Ø PCMGS tubing, Male Luer with ¹/₄" barb, cap, slide clamp - Port 1: Break-off injection site
Port 2: Female MPC connector with ¹/₄ Barb, Male MPC plug
Port 3: Male MPC connector with ¹/₄ Barb, Female MPC cap
Spezifikationen
Zugehörige Informationen
Frequently Bought Together