
Trypan blue solution 0,4% (w/v) in PBS stain for histology pH 7,5 ±0,5, Corning®
Catalog # MDTC25-900-CI
Supplier: Corning
CAS Number:
Specifications
- Pk:1
- Seq:2445
- Pk Info:1×100 ml
Specifications
About this item
:
:960.82 g/mol
:GHS08
Storage Temperature:Ambient
CAS Number:72-57-1
Formula:C₃₄H₂₄N₆Na₄O₁₄S₄
MDL Number:MFCD00003969