Interior and surface cleaners, SURE®
Catalog # 115-2961
Supplier: Diversey
Specifications
- Description:Interior and surface cleaner, 2×5 L
- Seq:02
- Environmentally Preferable:Sustainable Materials,Safer Human and Environmental Health,Sustainable Packaging
- Pk:2 Bottle
Specifications
About this item
Concentrated all-purpose detergent for cleaning of hard surfaces, such as floors, walls, doors, ceilings and general equipment. Can also be used in food premises.
- Suitable for cleaning of all water resistant hard surfaces, including aluminium and other soft metals
- Effective in all water conditions
- No fragrance or dye
- Contains plant-based ingredients from renewable sources decomposed by natural processes
- No environmental hazard classifications, no safety warnings for user
- Concentrated formulation gives excellent economy of use
- Fast acting, fast drying
SURE® interior and surface cleaner is part of a unique range of plant-based, 100% biodegradable cleaning products that deliver the professional cleaning results you need - while being safe for people and kind to the environment. SURE® natural cleaning products provide safety and sustainability without compromising on performance.
Certifications: EU-Ecolabel (EU:NL/020/045).
The surfactants used in this product are in compliance with the requirement of EU Detergent Regulation, EC 648/2004.
The surfactants used in this product are in compliance with the requirement of EU Detergent Regulation, EC 648/2004.