ContactUsLinkComponent
Scissors, duck, Rubis®
Catalog # 233-0224
Supplier:  OUTILS RUBIS
undefined
Scissors, duck, Rubis®
Catalog # 233-0224
Supplier:  OUTILS RUBIS
Specifications
  • Type:
    Straight, blunt/blunt
  • Length:
    92 mm
  • Pk:
    1
  • Tip size:
    1 mm
Specifications
About this item
Micro scissors with beak-shaped tips.
  • Produced from hardened stainless steel AISI420
  • Highly resistant against acids and alkaline
  • With ergonomically designed Rubis Twist® grips
  • No Medical Device (MD) product
Made in Switzerland to the highest quality standards. Rubis products are the result of traditional craftsmanship, strict throughput controls and a finishing process under the magnifying glass. No tool leaves the factory without having passed the strict quality controls. Neither scratches nor inaccuracies in material or surface are permitted.