Order Entry
Austria
ContactUsLinkComponent
 
Newborn bovine calf serum, New Zealand, HyClone™
 
undefined
Newborn bovine calf serum, New Zealand, HyClone™

 

  • Catalog No:
  • SH30401.01IR25
  • SH30401.02IR30
  • SH30401.02-45
  • SH30401.02IR25
  • SH30401.01-45
  • SH30401.01IR30
  • Origin:
  • Animal
  • Animal
  • Animal
  • Animal
  • Animal
  • Animal
  • Endotoxin:
  • ≤25 EU/ml
  • ≤25 EU/ml
  • ≤25 EU/ml
  • ≤25 EU/ml
  • ≤25 EU/ml
  • ≤25 EU/ml
  • Haemoglobin:
  • ≤30 mg/dl
  • ≤30 mg/dl
  • ≤30 mg/dl
  • ≤30 mg/dl
  • ≤30 mg/dl
  • ≤30 mg/dl
  • Storage temperature:
  • Sera should be stored at −10 °C or lower. Once thawed, sera should be stored at 2...8 °C.
  • Sera should be stored at −10 °C or lower. Once thawed, sera should be stored at 2...8 °C.
  • Sera should be stored at −10 °C or lower. Once thawed, sera should be stored at 2...8 °C.
  • Sera should be stored at −10 °C or lower. Once thawed, sera should be stored at 2...8 °C.
  • Sera should be stored at −10 °C or lower. Once thawed, sera should be stored at 2...8 °C.
  • Sera should be stored at −10 °C or lower. Once thawed, sera should be stored at 2...8 °C.

 

Frequently Bought Together