Orders LinkContactUsLinkComponent
Sludge Judge® sampling tools
Sludge Judge® sampling tools
Sludge Judge® sampling tools
Sludge Judge® sampling tools
Sludge Judge® sampling tools
The Sludge Judge line of collection tools are dependable and long-lasting.
  • Designed to take accurate readings of settleable solids
  • Ideal for anywhere that accurate sample levels of settleable solids in noncaustic materials are needed
  • Maximum strength and rigidity
  • Treated to reduce deterioration from sun's harmful rays
  • Durable in cold temperature
  • Float valve for easy sample collection and release
Do NOT use for sampling ethanol or ethanol blended fuel. One-year warranty.
To use:
1. Slowly lower the Sludge Judge® to the bottom of the tank. The float valve opens, allowing materials to flow in. When the bottom has been reached and the pipe has filled to the surface level, tug slightly on the rope as the unit is raised. This sets the check valve, trapping the mixture inside.
2. When the unit has been raised clear of the liquid, the amount of solids in the sample can be read using the 0,31 m increments marked on the pipe sections. To release the material in the unit, touch the pin extending from the bottom section against a hard surface. This opens the check valve to drain the sample.
Dokumente
Product Family Options
  • Description
    Capacity
    Length
    Ø
  • Sampler, Sludge Judge®, complete unit
    89 ml/0,31 m
    4,60 m (3×1,53 m)
    19,0 mm
    1 * 1 ST
    1 * 1 ST - 492,00 €
    492,00 €
    1 * 1 ST
       492,00 €
  • Sampler, Sludge Judge® II, complete unit
    178 ml/0,31 m
    4,60 m (3×1,53 m)
    31,8 mm
    1 * 1 ST
    1 * 1 ST - 715,00 €
    715,00 €
    1 * 1 ST
       715,00 €
Accessories
  •  
    Description
  • NASCB01229WA
    Sludge nabber, up to 200 cm
    1 * 1 ST - 339,00 €
  •  
    Description
  • NASCB01291WA
    Cleaning brush for Sludge Judge and Sludge Judge Ultra, 183 cm
    1 * 1 ST - 151,00 €
  •  
    Description
  • NASCB01310WA
    Swing sampler, up to 365 cm
    1 * 1 ST - 623,00 €
  •  
    Description
  • NASCB01366WA
    Swing sampler, up to 730 cm
    1 * 1 ST - 799,00 €
  •  
    Description
  • NASCC09249WA
    Carrying bag for any Sludge Judge
    1 * 1 ST - 180,00 €
  •  
    Description
  • NASCC09250WA
    Sludge Judge bottom section, 152 cm
    1 * 1 ST - 280,00 €
  •  
    Description
  • NASCC09251WA
    Sludge Judge extension section, 152 cm
    1 * 1 ST - 179,00 €