Bench top biohazard bag holder kit, SP Bel-Art Poxygrid®
Economical and convenient, this bench top disposal system is perfect for collecting pipette tips, microcentrifuge tubes, etc during daily use.
- Snug fitting polypropylene plastic cover helps hold the bag securely in place
- Reusable, can be cleaned by hand or in automated dish washers
- Autoclavable
The rectangular steel wire frame holds a 220×280 mm polyethylene bag which is coated in easy to clean, durable, orange colour epoxy.
: Cover provides compliance with 29 CFR 1910.1030(d)(2)(xiii)(A), which requires a cover on any container used to store, transport, or ship biohazardous wastes.
: Kit includes 100 bench top biohazard bags (129-4922); can also be used with Clavies autoclavable bags.
Frequently Bought Together