Industrial Gloves, ActivArmr® 28-360
Supplier: Ansell
Certificates
About this item
The ActivArmr® range of protective gloves combines the outstanding protection of Kevlar®-reinforced construction with the grip and flexibility of Ansell’s advanced nitrile coating technology.
- Heavy weight design provides feeling of cushioning for the wearer
- NitraGold™ cuffs provide excellent protection
- Outstanding mechanical protection against cuts and abrasion
Ideal for safe handling of oily heavy metal parts, especially in the automotive area.
Certifications: EN 388:2016 (3321B), EN 407 (X1XXXX); Cat. III
Specifications
- Catalog No:
- ANSE28-360/10
- ANSE28-360/8
- ANSE28-360/9
- Model:
- 28-360
- 28-360
- 28-360
- Ambidextrous / hand-specific:
- Hand-specific
- Hand-specific
- Hand-specific
- Disposable / reusable:
- Reusable
- Reusable
- Reusable
- Liner material:
- Kevlar®/cotton
- Kevlar®/cotton
- Kevlar®/cotton
- Finish:
- Full coating (foam nitrile)
- Full coating (foam nitrile)
- Full coating (foam nitrile)
- Cuff style:
- Safety cuff
- Safety cuff
- Safety cuff
- EN norm:
- EN 388:2016 (3321B), EN 407 (X1XXXX)
- EN 388:2016 (3321B), EN 407 (X1XXXX)
- EN 388:2016 (3321B), EN 407 (X1XXXX)
- Anti-static:
- +
- +
- +