Flow Cell Priming Kit
Supplier: Oxford Nanopore Technologies
Certificates
About this item
Additional flow cell priming reagents to enable additional loading of more library during a sequencing run.
- The kit can be used to prime a MinION™ Flow Cell for sequencing or to add more fuel to a flow cell during a sequencing experiment.
Flow Cell Priming Kit V14 (EXP-FLP004) ― the Flow Cell Priming Kit V14 contains Flow Cell Flush (FCF) and Flow Cell Tether (FCT), sufficient for 6 reactions. This kit is compatible with the latest library preparation kit chemistry (V14) and R10.4.1 Flow Cells.
Note: This kit is not compatible with the Ultra-Long DNA Sequencing Kit V14, which requires specific flow cell priming reagents.
Delivery: Kits are shipped at 2 to 8 °C in an environmentally-friendly temperature-controlled shipping box. Upon receipt, please place the product in a long-term storage location at −20 °C. Oxford Nanopore Technologies deem the useful life of the product to be 3 months from receipt by the customer.
Caution: Oxford Nanopore Technologies products are not intended for use for health assessment or to diagnose, treat, mitigate, cure, or prevent any disease or condition.
Frequently Bought Together