Orders LinkContactUsLinkComponent
VWR®, Solid Euro Containers
Supplier:   VWR Collection
VWR®, Solid Euro Containers
Supplier:  VWR Collection
Certificates
VWR®, Solid Euro Containers
VWR®, Solid Euro Containers
16 Options Available Below
Product Details & Documents
These plastic stackable Euro containers are ideal for simple storage solutions as well as highly automated systems. Made from strong plastic these containers are ideal for transporting goods and food safely. They are very stable when loaded due to reinforced frame and corner construction.
  • Made of PPC (polypropylene copolymer)
  • Suitable for every industry and for any application
  • Smooth surfaces make them easy to clean
  • Ergonomic handles for effective manual handling
  • Suitable for –20 to +65 °C storage
  • 100% recyclable
The solid Euro containers are available in universal Euros sizes: 300×200, 400×300, 600×400 or 800×600 mm. They are perfect for palletisation. They are also suitable for manual and automatic conveyor handling. The standard colour is grey.
 : Guaranteed food-safe.
 : Trolleys and lids are optional and can be supplied for the entire range.
Product Family Options
Containers
Product Information
  • Capacity
    W×D×H ext.
    W×D×H int.
    Availability
    Price
  • 216-2390
    Solid Euro container, 15 kg load capacity, closed handles
    Solid Euro container, 15 kg load capacity, closed handles
    2,9 L
    300×200×70 mm
    268×168×68 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 8,60 €
    8,60 €
    1 * 1 ST
       8,60 €
  • 216-2392
    Solid Euro container, 25 kg load capacity, closed handles
    Solid Euro container, 25 kg load capacity, closed handles
    5,4 L
    400×300×70 mm
    367×268×58 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 11,00 €
    11,00 €
    1 * 1 ST
       11,00 €
  • 216-2391
    Solid Euro container, 15 kg load capacity, closed handles
    Solid Euro container, 15 kg load capacity, closed handles
    7,2 L
    300×200×170 mm
    268×168×168 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 10,30 €
    10,30 €
    1 * 1 ST
       10,30 €
  • 216-2393
    Solid Euro container, 25 kg load capacity, closed handles
    Solid Euro container, 25 kg load capacity, closed handles
    11,2 L
    400×300×120 mm
    367×268×117 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 12,70 €
    12,70 €
    1 * 1 ST
       12,70 €
  • 216-2398
    Solid Euro container, 25 kg load capacity, closed handles
    Solid Euro container, 25 kg load capacity, closed handles
    13,7 L
    600×400×70 mm
    567×367×67 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 15,80 €
    15,80 €
    1 * 1 ST
       15,80 €
  • 216-2394
    Solid Euro container, 25 kg load capacity, open handles
    Solid Euro container, 25 kg load capacity, open handles
    15,9 L
    400×300×170 mm
    367×268×167 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 14,90 €
    14,90 €
    1 * 1 ST
       14,90 €
  • 216-2395
    Solid Euro container, 25 kg load capacity, open handles
    Solid Euro container, 25 kg load capacity, open handles
    20,8 L
    400×300×220 mm
    367×268×217 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 16,60 €
    16,60 €
    1 * 1 ST
       16,60 €
  • 216-2399
    Solid Euro container, 25 kg load capacity, closed handles
    Solid Euro container, 25 kg load capacity, closed handles
    23,9 L
    600×400×120 mm
    567×367×117 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 19,00 €
    19,00 €
    1 * 1 ST
       19,00 €
  • 216-2396
    Solid Euro container, 25 kg load capacity, open handles
    Solid Euro container, 25 kg load capacity, open handles
    25,6 L
    400×300×270 mm
    367×268×267 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 19,50 €
    19,50 €
    1 * 1 ST
       19,50 €
  • 216-2397
    Solid Euro container, 25 kg load capacity, open handles
    Solid Euro container, 25 kg load capacity, open handles
    30 L
    400×300×320 mm
    367×268×317 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 21,30 €
    21,30 €
    1 * 1 ST
       21,30 €
  • 216-2400
    Solid Euro container, 25 kg load capacity, open handles
    Solid Euro container, 25 kg load capacity, open handles
    35,6 L
    600×400×170 mm
    567×367×167 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 22,10 €
    22,10 €
    1 * 1 ST
       22,10 €
  • 216-2401
    Solid Euro container, 25 kg load capacity, open handles
    Solid Euro container, 25 kg load capacity, open handles
    44,2 L
    600×400×220 mm
    567×367×217 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 25,10 €
    25,10 €
    1 * 1 ST
       25,10 €
  • 216-2402
    Solid Euro container, 25 kg load capacity, open handles
    Solid Euro container, 25 kg load capacity, open handles
    54,4 L
    600×400×270 mm
    567×367×267 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 29,80 €
    29,80 €
    1 * 1 ST
       29,80 €
  • 216-2409
    Solid Euro container, 25 kg load capacity, open handles
    Solid Euro container, 25 kg load capacity, open handles
    64,5 L
    600×400×320 mm
    567×267×317 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 31,50 €
    31,50 €
    1 * 1 ST
       31,50 €
  • 216-2410
    Solid Euro container, 35 kg load capacity, closed handles
    Solid Euro container, 35 kg load capacity, closed handles
    91 L
    800×600×220 mm
    767×567×216 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 62,60 €
    62,60 €
    1 * 1 ST
       62,60 €
  • 216-2416
    Solid Euro container, 35 kg load capacity, closed handles
    Solid Euro container, 35 kg load capacity, closed handles
    168 L
    800×600×420 mm
    767×567×416 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 96,60 €
    96,60 €
    1 * 1 ST
       96,60 €
Lids
Product Information
  • Catalog #
    Description
    Dimensions
    Availability
    Price
  • Lid 300×200, 80 kg load capacity
    300×200×10 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 8,00 €
    8,00 €
    1 * 1 ST
       8,00 €
Product Information
  • Catalog #
    Description
    Dimensions
    Availability
    Price
  • Lid 800×600, 80 kg load capacity
    800×400×10 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 38,50 €
    38,50 €
    1 * 1 ST
       38,50 €
Product Information
  • Catalog #
    Description
    Dimensions
    Availability
    Price
  • Lid 400×300, 80 kg load capacity
    400×300×10 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 8,70 €
    8,70 €
    1 * 1 ST
       8,70 €
Product Information
  • Catalog #
    Description
    Dimensions
    Availability
    Price
  • Lid 600×400, 80 kg load capacity
    600×400×10 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 11,30 €
    11,30 €
    1 * 1 ST
       11,30 €