ContactUsLinkComponent
GLS 80® wide mouth connection transfer cap system, DURAN®
GLS 80® wide mouth connection transfer cap system, DURAN®
  217-0896
undefined
GLS 80® wide mouth connection transfer cap system, DURAN®
  217-0896
  • Description:
    Screw cap GLS 80, 4 ports with GL 18 thread, EPDM seal
  • Seq:
    03
  • Pk:
    2
PP and PTFE.
  • Flexible modular system
  • Very good temperature resistance
  • Safe transfer of liquid media within a closed and sterile system (evaporation is reduced)
Screw Cap GLS 80 with four ports (GL thread). Five different hose diameters (3,2; 6,0; 8,0; 10,00 and 12,0 mm) can be connected. Sterile pressure equalisation is possible through use of membrane filter. Unused ports can be provided with a blind cap.